Skip to Content
Merck
All Photos(2)

Key Documents

B7906

Sigma-Aldrich

1-Butanol

for molecular biology, ≥99%

Synonym(s):

n-Butanol, Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
969148
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

vapor density

2.55 (vs air)

vapor pressure

5 (4 mmHg) at 20 °C hPa

Assay

≥99%

form

liquid

autoignition temp.

649 °F

expl. lim.

11.2 %

color

colorless

refractive index

n20/D 1.399 (lit.)

pH

7 (20 °C, 70 g/L)

bp

116-118 °C (lit.)

mp

−90 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

CCCCO

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butanol is a clear, colorless alcohol that can be used for the removal of ethidium bromide from DNA purified by CsCl gradient ultracentrifugation. It is also suitable for use in the concentration of dilute nucleic acid solutions by repeated extractions, thus improving recovery by ethanol precipitation.

Application

1-Butanol has been used:
  • in the derivatization process for measuring methylated arginine levels in blood samples
  • in liquid-chromatography mass spectrometry (LC-MS) analysis of lysine modifications by deacetylase CobB
  • in the extraction of plant material from Vigna unguiculata
  • to extract and purify amplified human DNA samples before further amplification by PCR followed by sequencing

1-Butanol is suitable for use in nucleic acid purification and concentration protocols.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietro Benettoni et al.
Biointerphases, 15(2), 021005-021005 (2020-03-28)
Nowadays, high-resolution imaging techniques are extensively applied in a complementary way to gain insights into complex phenomena. For a truly complementary analytical approach, a common sample carrier is required that is suitable for the different preparation methods necessary for each
Hon Sing Leong et al.
Nature protocols, 5(8), 1406-1417 (2010-07-31)
Viral nanoparticles are a novel class of biomolecular agents that take advantage of the natural circulatory and targeting properties of viruses to allow the development of therapeutics, vaccines and imaging tools. We have developed a multivalent nanoparticle platform based on
Elena Castellanos-Rizaldos et al.
PloS one, 7(12), e51362-e51362 (2012-12-14)
Multiplex detection of low-level mutant alleles in the presence of wild-type DNA would be useful for several fields of medicine including cancer, pre-natal diagnosis and infectious diseases. COLD-PCR is a recently developed method that enriches low-level mutations during PCR cycling
Mehmet Nuri Atalar et al.
Life sciences, 231, 116577-116577 (2019-06-19)
Galectin-3 is associated with the process of inflammation and fibrosis. The aim of this study was both to evaluate of galectin-3, methylated arginines and hs-CRP in subjects with type 2 diabetes and prediabetes and to investigate a relation between serum
Laura Broutier et al.
Nature protocols, 11(9), 1724-1743 (2016-08-26)
Adult somatic tissues have proven difficult to expand in vitro, largely because of the complexity of recreating appropriate environmental signals in culture. We have overcome this problem recently and developed culture conditions for adult stem cells that allow the long-term

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service