Merck
Hydrophobic properties of alkaline phosphatases.
The International journal of biochemistry (1986-01-01)
R W Wulkan, M I Huijskes-Heins, B Leijnse
ABSTRACT
The butanol extraction method of Morton (1950), a routine step in enzyme purification, is discussed with special reference to a hydrophobic form of alkaline phosphatase from human liver tissue. This form slowly precipitates from butanol-extracted liver tissue homogenates stored at 4 degrees C. Furthermore, it is lost when acetone precipitation is applied as a purification procedure. The soluble form in liver tissue is shown to have a higher relative hydrophobicity than the serum liver/bone isoenzyme. The use of sodium cholate in the isolation of the hydrophobic form produces an artefact in isoelectric focusing, which can be abolished by dialysis prior to focusing.
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
1-Butanol, suitable for HPLC
Sigma-Aldrich
1-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
1-Butanol, SAJ first grade, ≥99.0%
Supelco
1-Butanol, suitable for HPLC, 99.8%
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%