Skip to Content
Merck
All Photos(2)

Documents

1.06393

Supelco

Sodium carbonate

anhydrous, for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

anhydrous soda, Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
MDL number:
UNSPSC Code:
12352302
EC Index Number:
207-838-8
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.9% dry basis (acidimetric)

form

powder

potency

4090 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

loss

≤1% loss on drying, 300 °C

pH

11.16 (25 °C, 4 g/L in H2O)

mp

851 °C (lit.)

density

2.53 g/cm3 at 20 °C

bulk density

1100 kg/m3

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
silicate (as SiO2): ≤0.005%
sulfur compounds (as SO42-): ≤0.003%

cation traces

Al: ≤0.001%
As: ≤0.0001%
Ca: ≤0.005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
N: ≤0.001%
heavy metals: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Linkage

Replaces: SX0400-20; SX0400-5

Analysis Note

Assay (acidimetric; dried substance): ≥ 99.9 %
Insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.001 %
Silicate (as SiO₂): ≤ 0.005 %
Total sulfur (as SO₄): ≤ 0.003 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (ACS): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.001 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.0005 %
Loss on drying (300 °C): ≤ 1.0 %
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.

A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.

A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.

A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service