Skip to Content
Merck
All Photos(1)

Key Documents

1.06516

Supelco

Sodium thiosulfate pentahydrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Sodium thiosulfate pentahydrate, Antichlor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O3S2 · 5H2O
CAS Number:
Molecular Weight:
248.18
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-867-5
NACRES:
NA.21
Assay:
99.5-101.0% (iodometric)
grade:
ACS reagent
form:
solid
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.
suitable for SM 2310
suitable for SM 2320
suitable for SM 4500 - NH3

product line

EMSURE®

Assay

99.5-101.0% (iodometric)

form

solid

potency

>5000 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

pH

6.0-7.5 (20 °C, 100 g/L in H2O)

mp

48 °C

density

1.74 g/cm3 at 20 °C

bulk density

1000 kg/m3

anion traces

chloride (Cl-): ≤0.008%
sulfate, sulfite (as SO42-): ≤0.1%
sulfide (S2-): passes test

cation traces

Ca: ≤0.002%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.001%
Mg: ≤0.001%
N: ≤0.002%
Pb: ≤0.0005%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[S](=S)(=O)([O-])[O-].O.O.O.O.O

InChI

1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2

InChI key

PODWXQQNRWNDGD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Synthesis of Aryl Amides from Acyl-Bunte Salts and Aryl Azides: Details the application of sodium thiosulfate pentahydrate in synthetic organic chemistry, specifically in the synthesis of aryl amides, illustrating its utility in facilitating complex chemical reactions (Chen et al., 2023).

Linkage

Replaces: SX0815-1; SX0815

Analysis Note

Assay (iodometric): 99.5 - 101.0 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water, 25 °C): 6.0 - 7.5
pH-value (10 %; water): 6.0 - 7.5
Chloride (Cl): ≤ 0.008 %
Sulfate and sulphite (as SO₄): ≤ 0.1 %
Sulfide (S): passes test
Heavy metals (as Pb): ≤ 0.001 %
Total nitrogen (N): ≤ 0.002 %
Ca (Calcium): ≤ 0.002 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.001 %
Mg (Magnesium): ≤ 0.001 %
Pb (Lead): ≤ 0.0005 %
Corresponds to ACS, ISO, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service