Skip to Content
Merck
All Photos(4)

Key Documents

222321

Sigma-Aldrich

Sodium carbonate

ACS reagent, anhydrous, ≥99.5%, powder or granules

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
powder or granules
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous

Quality Level

Agency

suitable for SM 4500 - NH3

Assay

≥99.5%

form

powder or granules

impurities

≤0.005% silica
≤0.01% insolubles

loss

≤1.0% loss on heating, 285 °C

pH

12 (25 °C, 106 g/L)

mp

851 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
sulfur compounds (as SO42-): ≤0.003%

cation traces

Ca: ≤0.03%
Fe: ≤5 ppm
K: ≤0.005%
Mg: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium carbonate (Soda) is a white salt. It can be synthesized by the electrolysis of the molten sodium hydroxide or sodium chloride.[1]
Sodium carbonate is a water soluble inorganic salt commonly used as a weak base.[2] Its aqueous solution has the ability to uptake carbon dioxide.[3] It can also catalyze the conversion of sewage sludge to liquid fuels.[4]

Application

Sodium carbonate may be used as a reducing agent for the preparation of graphene sheets with good electrocapacity from graphene oxide dispersion.[5]
It may also be used in procedures for the synthesis of the following:
  • Biaryls via palladium-catalyzed cross-coupling reaction of phenylboronicacid with haloarenes.[6]
  • Chalcones and azachalcones via aldol condensation of aldehydes with ketones in aqueous medium.[7]
  • Enynes via copper-catalyzed allylation of functionalized alkynes.[2]
Sodium carbonate may be used in the following studies:
  • Determination of total polyphenolic index of wine products from grape by the Folin-Ciocalteu method.[8]
  • To compose the ELISA (Enzyme-Linked Immunosorbent Assay) coating buffer.[9]
  • Preparation of the antibody coating buffer for use in IGF-I kinase receptor activation assay (KIRA) assay.[10]

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Absorption of carbon dioxide into aqueous sodium hydroxide and sodium carbonate-bicarbonate solutions.
Hikita H, et al.
Chemical Engineering Journal, 11(2), 131-141 (1976)
Efficient and clean aldol condensation catalyzed by sodium carbonate in water.
Zhang Z, et al.
Chemistry Letters (Jpn), 32(10), 966-967 (2003)
A green and efficient method to produce graphene for electrochemical capacitors from graphene oxide using sodium carbonate as a reducing agent.
Jin Y, et al.
Applied Surface Science, 268, 541-546 (2013)
Liquid fuel production from sewage sludge by catalytic conversion using sodium carbonate.
Yokoyama S-Y, et al.
Fuel: The Science and Technology of Fuel and Energy, 66(8), 1150-1155 (1987)
Copper catalyzed regioselective coupling of allylic halides and alkynes promoted by weak inorganic bases.
Bieber LW and da Silva MF
Tetrahedron Letters, 48(40) , 7088-7090 (2007)

Articles

Development and validation of a fatty acid methyl esterification method, using methanolic HCl reagent, for FAME analysis by GC of vegetable oils and fats.

Development and validation of a fatty acid methyl esterification method, using methanolic HCl reagent, for FAME analysis by GC of vegetable oils and fats.

Development and validation of a fatty acid methyl esterification method, using methanolic HCl reagent, for FAME analysis by GC of vegetable oils and fats.

Development and validation of a fatty acid methyl esterification method, using methanolic HCl reagent, for FAME analysis by GC of vegetable oils and fats.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service