Skip to Content
Merck
All Photos(2)

Key Documents

1.06649

Supelco

Sodium sulfate

anhydrous for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-820-9
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (alkalimetric)
98.5-101% dry basis (alkalimetric)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

loss

≤0.5% loss on drying, 130 °C
≤0.5% loss on ignition, 800 °C

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

bulk density

1400‑1600 kg/m3

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%

cation traces

As: ≤0.0001%
Ca: ≤0.005%
Fe: ≤0.0005%
K: ≤0.002%
Mg: ≤0.001%
N: ≤0.0005%
heavy metals: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Linkage

Replaces: SX0761-20

Analysis Note

Assay (alkalimetric): ≥ 99.0 %
Assay (alkalimetric, calculated on dried substance): 98.5 - 101.0 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.01 %
Acidity or alkalinity: passes test
pH-value (5 %; water; 25 °C): 5.2 - 8.0
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.001 %
Total nitrogen (N): ≤ 0.0005 %
Heavy metals (ACS): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.002 %
Mg (Magnesium): ≤ 0.001 %
Loss on drying (130 °C): ≤ 0.5 %
Loss on ignition (800 °C): ≤ 0.5 %
Corresponds to ACS, ISO, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Method: Photometric determination with 3,3'-Diaminobenzidine

Spectrophotometric determination of proline by the Ninhydrin method, per EN 1141 and German Food and Feed Code §64 LFBG 31.00-7.

Spectrophotometric determination of proline by the Ninhydrin method, per EN 1141 and German Food and Feed Code §64 LFBG 31.00-7.

Spectrophotometric determination of proline by the Ninhydrin method, per EN 1141 and German Food and Feed Code §64 LFBG 31.00-7.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service