Skip to Content
Merck
All Photos(1)

Documents

B-903

Supelco

Bromazepam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10BrN3O
CAS Number:
Molecular Weight:
316.15
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

Brc1ccc2NC(=O)CN=C(c3ccccn3)c2c1

InChI

1S/C14H10BrN3O/c15-9-4-5-11-10(7-9)14(17-8-13(19)18-11)12-3-1-2-6-16-12/h1-7H,8H2,(H,18,19)

InChI key

VMIYHDSEFNYJSL-UHFFFAOYSA-N

Gene Information

human ... GABRA2(2555)

General description

A Certified Snap-N-Spike® Solution suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, and urine drug testing. Bromazepam is a benzodiazepine used for the treatment of anxiety.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marlo Cunha et al.
Arquivos de neuro-psiquiatria, 67(2A), 214-218 (2009-06-24)
This study aimed to elucidate cortical mechanisms and to identify the areas where occur such mechanisms due to interaction between bromazepam and motor learning. The sample was composed of 45 healthy subjects randomly distributed in 3 groups: placebo (n=15), bromazepam
Megadose bromazepam and zolpidem dependence: two case reports of treatment with flumazenil and valproate.
Gianluca Quaglio et al.
Substance abuse, 33(2), 195-198 (2012-04-12)
M S Attia
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(4), 972-976 (2009-09-29)
A rapid, simple and sensitive spectrofluorimetric method for determination of trace amount of bromazepam is developed. In phosphate buffer of pH 7.4. The bromazepam enhance the luminescence intensity of the Eu(3+) ion in Eu(3+)-bromazepam complex at lambda(ex)=390nm. The produced luminescence
Yvan Gaillard et al.
Forensic science international, 207(1-3), e16-e18 (2011-01-05)
We reported on the death by poisoning of a one-month-old baby that had followed the death of one of her sister (due to cyamemazine overdose). Exhumation of the corpse was done 8 months after burial and revealed the presence of
A rare case of male hysteria - hemiparesis successfully treated with amitriptyline.
Cedo D Miljevic et al.
Psychiatria Danubina, 23(3), 281-284 (2011-10-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service