X1202
m-Xylylenediamine
99%
Synonym(s):
1,3-Benzenedimethanamine, m-XDA, 1,3-Bis(aminomethyl)benzene, α,α′-Diamino-m-xylene
About This Item
Recommended Products
vapor pressure
15 mmHg ( 145 °C)
Quality Level
Assay
99%
refractive index
n20/D 1.571 (lit.)
bp
265 °C/745 mmHg (lit.)
density
1.032 g/mL at 25 °C (lit.)
SMILES string
NCc1cccc(CN)c1
InChI
1S/C8H12N2/c9-5-7-2-1-3-8(4-7)6-10/h1-4H,5-6,9-10H2
InChI key
FDLQZKYLHJJBHD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- As a monomer for the synthesis of poly(vinylogous urethane) vitrimers.
- As a starting material for the synthesis of carbamate-protected multisubstituted bis-guanidine.
- For the synthesis of polyamides by catalytic dehydrogenation with diols.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service