Skip to Content
Merck
All Photos(2)

Key Documents

369500

Sigma-Aldrich

4,4′-Methylenebis(2-methylcyclohexylamine), mixture of isomers

99%

Synonym(s):

2,2′-Dimethyl-4,4′-methylenebis(cyclohexylamine), 3,3′-Dimethyl-4,4′-diaminodicyclohexylmethane, 4,4′-Diamino-3,3′-dimethyldicyclohexylmethane, Bis(3-methyl-4-aminocyclohexyl)methane, DMDC, Dimethyldicykan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[C6H9(CH3)NH2]2
CAS Number:
Molecular Weight:
238.41
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

93-100 °C (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

CC1CC(CCC1N)CC2CCC(N)C(C)C2

InChI

1S/C15H30N2/c1-10-7-12(3-5-14(10)16)9-13-4-6-15(17)11(2)8-13/h10-15H,3-9,16-17H2,1-2H3

InChI key

IGSBHTZEJMPDSZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Methylenebis(2-methylcyclohexylamine) is an alicyclic diamine.

Application

4,4′-Methylenebis(2-methylcyclohexylamine) (MMCA) may be used:
  • in the synthesis of fully aliphatic polyimides
  • in the synthesis of fully aliphatic polyimide-siloxanes (APISiO)
  • to prepare epoxy resin with trimethylolpropane triglycidyl ether, used for the characterization of the structural organization in dragline silk by transmission X-ray microscopy (STXM).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

343.4 °F - closed cup

Flash Point(C)

173 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ohshima et al.
The Journal of toxicological sciences, 11(2), 79-93 (1986-05-01)
Amine-curing agent for epoxy resin, bis(4-amino-3-methyl-cyclohexyl)methane (commercial name; Laromin C) has been suspected to have induced in the workers some toxic signs such as collagen disease like scleroderma or polymyositis. Subacute toxicity of this agent was studied in rats following
S Ohshima et al.
Sangyo igaku. Japanese journal of industrial health, 26(3), 197-204 (1984-05-01)
Amine-curing agent for epoxy resin, bis(4-amino-3-methylcyclohexyl)methane, has been suspected of inducing toxic symptoms in man which resemble collagen disease such as scleroderma or polymyositis. We studied subacute toxicity of this agent by repeated oral administrations to rats. The agent was
H Ishikawa et al.
Dermatologica, 161(3), 145-151 (1980-01-01)
The presence of sclerosis-inducing glycosaminoglycan in the skin was confirmed in an experimental skin sclerosis induced by a chemical compound. An experimental skin sclerosis was first produced in the mouse with bis(4-amino-3-methylcyclohexyl)methane. Out of glycosaminoglycans isolated from the slightly changed
Jake Brooks et al.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 62, 126555-126555 (2020-06-12)
Neuromelanin-pigmented neurons, which are highly susceptible to neurodegeneration in the Parkinson's disease substantia nigra, harbour elevated iron levels in the diseased state. Whilst it is widely believed that neuronal iron is stored in an inert, ferric form, perturbations to normal
Fully aliphatic polyimides from adamantane-based diamines for enhanced thermal stability, solubility, transparency, and low dielectric constant.
Mathews AS, et al.
Journal of Applied Polymer Science, 102(4), 3316-3326 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service