Skip to Content
Merck
All Photos(2)

Documents

32851

Sigma-Aldrich

trans-1,4-Diaminocyclohexane

≥98.0% (GC)

Synonym(s):

trans-1,4-Cyclohexanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(NH2)2
CAS Number:
Molecular Weight:
114.19
Beilstein:
2801657
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

18 mmHg ( 87.2 °C)

Assay

≥98.0% (GC)

autoignition temp.

680 °F

expl. lim.

6 %

bp

197 °C (lit.)

mp

68-72 °C (lit.)

SMILES string

N[C@H]1CC[C@H](N)CC1

InChI

1S/C6H14N2/c7-5-1-2-6(8)4-3-5/h5-6H,1-4,7-8H2/t5-,6-

InChI key

VKIRRGRTJUUZHS-IZLXSQMJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-1,4-Diaminocyclohexane was used in preparation of fully aliphatic polyimides. It was employed as the structure-directing agent in the synthesis of novel two-dimensional layered zinc phosphate.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Vianello et al.
Protein expression and purification, 15(2), 196-201 (1999-03-02)
A novel, simple, and rapid procedure for the purification of pea seedling amine oxidase is reported. The crude enzyme, obtained by ammonium sulfate fractionation, was purified in two steps: the first one by anion-exchange chromatography and the second one by
Hwan-Chul Yu et al.
Journal of nanoscience and nanotechnology, 11(7), 6141-6147 (2011-11-30)
Fully aliphatic polyimides (APIs) were prepared from rel-(1'R,3S5'S)-spiro[furan-3(2H),6'-[3]oxabicyclo[3.2.1]octane]-2,2',4',5(4H)-tetrone (DAn) as unsymmetrical spiro dianhydride, and either cis-trans-1,4-diaminocyclohexane (mix-DACH) or trans-1,4-diaminocyclohexane (trans-DACH) as diamine. Structure of all prepared monomers and polymers was confirmed via 1H-NMR and FT-IR. The solubility, optical transparency, and
Jana Kasparkova et al.
Biochemical pharmacology, 79(4), 552-564 (2009-09-29)
Earlier studies have described promising antitumor activity of a large-ring chelate complex [PtCl(2)(cis-1,4-DACH)] (DACH=diaminocyclohexane). Encouraging antitumor activity of this analogue of cisplatin prompted us to perform studies focused on the mechanistic basis of pharmacological effects of this complex. Four early
Mohammad S Ali et al.
Journal of inorganic biochemistry, 96(4), 452-456 (2003-09-19)
A series of new platinum(II) and platinum(IV) adducts of type [P(II)(cis-1,4-DACH)LCl]NO(3,) where cis-1,4-DACH=cis-1,4-diaminocyclohexane, and L=9-ethylguanine, 1-methylcytosine, adenine, adenosine, cytosine, cytidine, guanine, and [Pt(IV)(cis-1,4-DACH)Ltrans-(X)(2)Cl]NO(3), (where Y=hydroxo or acetato), were synthesized and characterized by elemental analysis, infrared spectroscopy, and 1H and 195Pt
Shalini Balakrishnan et al.
The Journal of organic chemistry, 72(25), 9834-9837 (2007-11-06)
To introduce chirality and functional groups adjacent to guanidiniums to modulate specificity and affinity in recognition, N,N'-bis(Boc)-alpha-guanidino acids were synthesized from alpha-amino acid methyl esters. Protected alpha-guanidino acids coupled to cyclohexylamine and trans-1,4-diaminocyclohexane in good yield and with retention of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service