Skip to Content
Merck
All Photos(3)

Documents

180467

Sigma-Aldrich

1,3-Cyclohexanebis(methylamine), mixture of isomers

greener alternative

98%

Synonym(s):

1,3-Bis(aminomethyl)cyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CH2NH2)2
CAS Number:
Molecular Weight:
142.24
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.493 (lit.)

density

0.945 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

NCC1CCCC(CN)C1

InChI

1S/C8H18N2/c9-5-7-2-1-3-8(4-7)6-10/h7-8H,1-6,9-10H2

InChI key

QLBRROYTTDFLDX-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

1,3-Cyclohexanebis(methylamine), mixture of isomers (1,3-BAC), is used as a cross-linking agent that can be used in the formation of fiber-based membranes for the separation of CO2. It can also be used in the synthesis of open-framework bimetallic phosphites for potential usage in molecular sieves and ion exchangers.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ishita Neogi et al.
ChemSusChem, 10(19), 3765-3772 (2017-07-29)
A new broadband-emitting 2 D hybrid organic-inorganic perovskite (CyBMA)PbBr
Alice Chen et al.
Biochimica et biophysica acta, 1589(2), 124-139 (2002-05-15)
Retrograde transport dependent on coat protein I (COPI) was impaired using two different approaches and the effects on the retrograde transport of protein toxins were investigated. One approach was to study ldlF cells that express a temperature-sensitive defect in the
Use of aminoglycoside antibiotics and related compounds to study ADP-ribosylation factor (ARF)/coatomer function in Golgi traffic.
R K Draper et al.
Methods in enzymology, 329, 372-379 (2001-02-24)
Separation of 1, 3-Cyclohexanebis (methylamine) Isomers by Template Synthesis of Open-Framework Bimetallic Phosphites
Chen G, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 635(9-10), 1463-1469 (2009)
Reversible diamine cross-linking of polyimide membranes
Powell CE, et al.
Journal of Membrane Science , 291(1-2), 199-209 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service