Skip to Content
Merck
All Photos(3)

Key Documents

I504

Sigma-Aldrich

2-Imidazolidinethione

98%

Synonym(s):

2-Mercaptoimidazoline, 2-Thioxoimidazolidine, N,N′-Ethylenethiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N2S
CAS Number:
Molecular Weight:
102.16
Beilstein:
106275
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

196-200 °C (lit.)

SMILES string

S=C1NCCN1

InChI

1S/C3H6N2S/c6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)

InChI key

PDQAZBWRQCGBEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • 2-Imidazolidinethione has been used as a source of nitrogen and sulfur in the preparation of heteroatom functionalized carbon dots with enhanced photoluminescence (PL) emissions.
  • It is a precursor for the synthesis of gel-polymer electrolyte based on cross-linked polyepichlorohydrin terpolymer (GECO), which can be used in lithium-sulfur batteries.
  • It is also used as a vulcanization accelerator in the preparation of styrene-butadiene rubber (SBR) and rubber composites.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prominence of fusion temperature and engineering heteroatoms on multifarious emissive shifts in carbon dots.
Velusamy, Jayaramakrishnan et al.
Journal of Colloid and Interface Science, 528, 237-247 (2018)
A method to improve the mechanical performance of styrene-butadiene rubber via vulcanization accelerator modified silica.
Zhong, Bangchao et al.
Composites Science and Technology, 117, 46-53 (2015)
A highly stretchable gel-polymer electrolyte for lithium-sulfur batteries.
Choudhury, Soumyadip et al.
Polymer, 112, 447-456 (2017)
D Boers et al.
Human & experimental toxicology, 27(9), 721-727 (2008-12-02)
We conducted a multicenter prospective study to assess the effects of occupational exposure to ethylenebisdithiocarbamate fungicides and/or other pesticides on self-reported asthma and asthmatic symptoms. This multicenter study was conducted among 248 workers exposed to pesticides and 231 non-exposed workers
Kevin Tran et al.
Food chemistry, 140(1-2), 340-342 (2013-04-13)
To response to the need for a rapid, cost-effective, eco-friendly and efficient extraction process, a sensitive method for the determination of ethylenethiourea (ETU) in food matrices by high-performance liquid chromatography-mass spectrometry (HPLC-LC/MS) was developed and validated. ETU was extracted from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service