Skip to Content
Merck
All Photos(1)

Key Documents

45531

Supelco

2-Imidazolidinethione

PESTANAL®, analytical standard

Synonym(s):

2-Thioxoimidazolidine, N,N′-Ethylenethiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N2S
CAS Number:
Molecular Weight:
102.16
Beilstein:
106275
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

196-200 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

S=C1NCCN1

InChI

1S/C3H6N2S/c6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)

InChI key

PDQAZBWRQCGBEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renato Polimanti et al.
Annals of human biology, 37(3), 440-450 (2010-04-10)
Study of the association between genetic variability and individual susceptibility can help to characterize occupational or environmental risks due to xenobiotics. This study evaluates the influence of genetic components and environmental factors in relation to pesticide exposure. The study population
Yanlei Huang et al.
Journal of pediatric surgery, 46(7), 1400-1405 (2011-07-19)
The aim of this study was to study the effect of vitamin A deficiency (VAD) on the embryological development of anorectal malformations (ARMs) and the enteric nervous system. Female Sprague-Dawley rats were divided into 3 groups: VAD group, normal group
Xinyu Dong et al.
Journal of environmental sciences (China), 23(1), 65-69 (2011-04-12)
The experimental study on ozonation of ethylenethiourea (ETU) is conducted. The reaction of gas-phase ETU with 0.63 x 10(-6) mol/L ozone is carried out in a 200-L reaction chamber. The secondary organic aerosol (SOA) resulted from the ozonation of gas-phase
Ethylene thiourea.
Report on carcinogens : carcinogen profiles, 12, 191-194 (2011-08-20)
Reiko Kaneko et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 53(1), 52-56 (2012-03-28)
An improved analysis method for 2-mercaptoimidazoline in rubber products containing chlorine was developed. 2-Mercaptoimidazoline (20 µg/mL) is detected by means of TLC with two developing solvents in the official method. But, this method is not quantitative. Instead, we employed HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service