Skip to Content
Merck
All Photos(1)

Key Documents

115169

Sigma-Aldrich

Tetramethylthiourea

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCSN(CH3)2
CAS Number:
Molecular Weight:
132.23
Beilstein:
1744916
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

245 °C (lit.)

mp

75-77 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)C(=S)N(C)C

InChI

1S/C5H12N2S/c1-6(2)5(8)7(3)4/h1-4H3

InChI key

MNOILHPDHOHILI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethylthiourea molecule shows crystallographic two-fold symmetry with weak hydrogen bond packing, which connects the molecules to form layers.

Application

Tetramethylthiourea forms BF3 adduct with tetramethylselenourea.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allen Mambanda et al.
Dalton transactions (Cambridge, England : 2003), 39(15), 3595-3608 (2010-04-01)
The rate of displacement of the aqua ligands by three neutral nucleophiles (Nu) of different steric demands, namely thiourea (tu), N,N'-dimethylthiourea (dmtu) and N,N,N',N'-tetramethylthiourea (tmtu) and an anionic nucleophile (I(-)) in complexes of the form [{Pt(H(2)O)}(2)(N,N,N',N'-tetrakis(2-pyridylmethyl)-N(CH(2))(n)N](CF(3)SO(3))(4), n = 2 (En);
Y Usson et al.
Journal of neurocytology, 17(5), 639-648 (1988-10-01)
The proliferation of Schwann cells in the sciatic nerve of chick was studied from day 11 to day 27 of development in control and thyroid-deficient embryos. Hypothyroidism was induced by tetramethylthiourea injection on days 8 and 19 of incubation. The
A Shukla et al.
Cancer research, 61(5), 1791-1795 (2001-03-31)
Crystalline silica has been classified as a group 1 human carcinogen in the lung. However, its mechanisms of action on pulmonary epithelial cells which give rise to lung cancers are unclear. Using a nontransformed alveolar type II epithelial cell line
Alexius Freyberger et al.
Toxicology, 217(2-3), 169-175 (2005-11-22)
N,N,N',N'-Tetramethylthiourea (TMTU) is a rat goitrogen inducing thyroid hyperplasia, hypertrophy, and tumor formation. Little is known about the exact underlying mechanism of action. As thyroid peroxidase (TPO) and type I iodothyronine deiodinase (ID-I) have been established as targets of goitrogenic
Jiqin Liu et al.
Molecular immunology, 45(15), 3984-3989 (2008-07-29)
N-Acetylcysteine (NAC) has been widely used as an antioxidant in research, however, it has also been found to reduce the binding of TNF to its receptor independent of its antioxidative role. In this study, we investigated the effect of NAC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service