Skip to Content
Merck
All Photos(3)

Key Documents

C4706

Sigma-Aldrich

Tris(2-carboxyethyl)phosphine hydrochloride

powder

Synonym(s):

TCEP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15O6P · HCl
CAS Number:
Molecular Weight:
286.65
Beilstein:
3724376
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

description

Protect from moisture

Quality Level

form

powder

reaction suitability

reagent type: reductant

color

white

mp

175-177 °C

solubility

H2O: 50 mg/mL

density

1.041 g/cm3 at 25 °C

storage temp.

2-8°C

SMILES string

Cl[H].OC(=O)CCP(CCC(O)=O)CCC(O)=O

InChI

1S/C9H15O6P.ClH/c10-7(11)1-4-16(5-2-8(12)13)6-3-9(14)15;/h1-6H2,(H,10,11)(H,12,13)(H,14,15);1H

InChI key

PBVAJRFEEOIAGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(2-carboxyethyl)phosphine hydrochloride (TCEP.HCl) is a non-volatile solid. It is a strong reducing agent.1 It can be synthesized by the acid hydrolysis of tris(2-cyanoethyl)phosphine in refluxing aqueous HCl.1 It has various biological applications such as in vitro and in vivo reduction of disulfide bonds in various peptides and proteins. TCEP is a useful chelating agent for various heavy metal ions as Zn(II), Cd(II), Pb(II), and Ni(II).

Application

Tris(2-carboxyethyl)phosphine hydrochloride (TCEP. HCl) can be used:
  • As a reducing agent for the reduction of sulfoxides, sulfonyl chlorides, N-oxides, and azides. It can also be used in azide-alkyne cycloaddition reaction in the presence of a copper catalyst.
  • To reduce disulfide bonds in various proteins.
  • As a reagent for the selective reduction of disulfides in water.
  • To remove ruthenium-derived metathesis catalysts via aqueous washing of a crude reaction mixture when it is basified.
  • As a reducing agent for the reduction of various alkyl disulfides such as trans-4,5-dihydroxy-1,2-dithiane.
Water soluble reagent, used for selective reduction of disulfides. More stable than DTT and useful in mass spectrometry applications.

Caution

Storage conditions:protect from moisture.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grubbs, R. H.
Org. React., 22, 123-123 (2004)
Selective reduction of disulfides by tris(2-carboxyethyl)phosphine
Burns, J. A., et al.
The Journal of Organic Chemistry, 56(8), 2648-2648 (1991)
Selective reduction of disulfides by tris (2-carboxyethyl) phosphine.
Burns JA, et al.
The Journal of Organic Chemistry, 56(8), 2648-2650 (1991)
Tris (2-carboxyethyl) phosphine Hydrochloride
Yost Julianne M et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-9 (2001)
Coordination properties of tris (2-carboxyethyl) phosphine, a newly introduced thiol reductant, and its oxide.
Krezel A, et al.
Inorganic Chemistry, 42(6), 1994-2003 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service