Skip to Content
Merck
All Photos(1)

Key Documents

E45309

Sigma-Aldrich

Ethyl phenylpropiolate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CCOOC2H5
CAS Number:
Molecular Weight:
174.20
Beilstein:
639637
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.552 (lit.)

bp

260-270 °C (lit.)

density

1.055 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C#Cc1ccccc1

InChI

1S/C11H10O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-7H,2H2,1H3

InChI key

ACJOYTKWHPEIHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K A Davidson et al.
The Journal of investigative dermatology, 79(6), 378-382 (1982-12-01)
Glucocorticoids (anti-inflammatory steroids) are very potent inhibitors of mouse skin tumor promotion induced by 12-O-tetradecanoylphorbol-13-acetate (TPA). This report describes a high-affinity, limited-capacity binding component which specifically interacts with glucocorticoids and which is identified as a glucocorticoid (GC) receptor present in
J E Paulsen
Carcinogenesis, 11(8), 1255-1258 (1990-08-01)
We examined the effects of retinoic acid (RA) on epidermal DNA synthesis, induced by 12-O-tetradecanoylphorbol-13-acetate (TPA), a strong tumor promoter; mezerein (MEZ), a strong second stage promoter or ethylphenylpropiolate (EPP), a weak tumor promoter. RA reduced the initial wave of
G S Cameron et al.
Cancer research, 51(20), 5642-5648 (1991-10-15)
The ability of the hyperplasiogenic irritant ethyl phenylpropiolate (EPP) to act as a tumor promoter in two-stage carcinogenesis and to stimulate cellular events commonly cited as markers of tumor promoter action was evaluated. Treatment of adult, inbred SENCAR (SSIN) mice
Ampai Panthong et al.
Journal of ethnopharmacology, 91(2-3), 237-242 (2004-05-04)
Methanolic extracts from the heart wood, stem bark, and stem wood of Ventilago harmandiana Pierre (Family Rhamnaceae) were assessed for anti-inflammatory effects using both acute and chronic inflammatory models. Analgesic and antipyretic activities of the extracts were also evaluated. It
P Claeson et al.
Planta medica, 62(3), 236-240 (1996-06-01)
The topical anti-inflammatory activity of three non-phenolic linear 1,7-diarylheptanoids, previously isolated from a Thai medicinal plant, Curcuma xanthorrhiza (Zingiberaceae) and four new semi-synthetic derivatives of the naturally occurring compounds were assessed in the murine model of ethyl phenylpropiolate-induced ear edema.

Articles

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service