Skip to Content
Merck
All Photos(2)

Key Documents

E46607

Sigma-Aldrich

Ethyl propiolate

99%

Synonym(s):

Ethyl acetylenecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCO2C2H5
CAS Number:
Molecular Weight:
98.10
Beilstein:
878250
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.412 (lit.)

bp

120 °C (lit.)

density

0.968 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C#C

InChI

1S/C5H6O2/c1-3-5(6)7-4-2/h1H,4H2,2H3

InChI key

FMVJYQGSRWVMQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Halogenated derivatives were used as substrates for direct, asymmetric alkynylation of cyclic ß-ketoesters using chiral phase-transfer catalysts. Also employed in a one-pot , four-component synthesis of benzene-1,2,3,5-tetracarboxylates promoted by Ph3P.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Morrison-Whittle et al.
Frontiers in microbiology, 9, 910-910 (2018-06-06)
The products of microbial metabolism form an integral part of human industry and have been shaped by evolutionary processes, accidentally and deliberately, for thousands of years. In the production of wine, a great many flavor and aroma compounds are produced
Thomas B Poulsen et al.
Journal of the American Chemical Society, 129(2), 441-449 (2007-01-11)
The first organocatalytic enantioselective direct alpha-alkynylation of beta-ketoesters and 3-acyl oxindoles is described. It is demonstrated that activated beta-halo-alkynes undergo nucleophilic acetylenic substitution catalyzed by chiral phase-transfer compounds to afford the alkynylated products in high yields and excellent enantioselectivities. The
Katie Parish-Virtue et al.
Food chemistry, 271, 747-752 (2018-09-22)
Sauvignon blanc grapes were exposed to an ultra-violet (UV) light source post-hand harvest (whole bunches) or post-machine harvest. The thiol precursors S-3-(hexan-1-ol)-l-cysteine (Cys-3MH) and S-3-(hexan-1-ol)-l-glutathione (GSH-3MH) were quantified in the juices before and after UV treatment. Results showed that irradiation
Mikhail V Dobrynin et al.
Carbohydrate polymers, 241, 116327-116327 (2020-06-09)
Hydrosilylation catalyzed by the rhodium(I) complex [Rh(acac)(CO)2] or platinum(0)-based Karstedt's catalyst was employed to combine hydrophilic propargylated hydroxyethyl cellulose and hydrophobic hydride-terminated polydimethylsiloxane to give polymer hybrid structures. The final polymers were characterized by FTIR, solid state 1H, 13C and
Helvetica Chimica Acta, 89, 2918-2918 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service