Skip to Content
Merck
All Photos(1)

Documents

281492

Sigma-Aldrich

Ethyl benzoylacetate

technical grade, 90%

Synonym(s):

Benzoylacetic acid ethyl ester, Ethyl 3-oxo-3-phenylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2COOC2H5
CAS Number:
Molecular Weight:
192.21
Beilstein:
389944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor density

6.6 (vs air)

Assay

90%

form

liquid

refractive index

n20/D 1.52 (lit.)

bp

265-270 °C (lit.)

solubility

alcohol: miscible
diethyl ether: miscible
water: insoluble

density

1.11 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1ccccc1

InChI

1S/C11H12O3/c1-2-14-11(13)8-10(12)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

GKKZMYDNDDMXSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl benzoylacetate is an ester. It undergoes microbial reduction by bakers′ yeast (Saccharomyces cerevisiae), Beauveria sulfurescens or Geotrichum candidum to afford ethyl (S)-3-hydroxy-3-phenylpropionate. It undergoes Claisen condensation reaction with malononitrile to afford 2-cyano-5-phenyl-3,5-dioxopentanonitrile which on cyclization followed by coupling with diazonium salts yields azo derivatives.

Application

Ethyl benzoylacetate was sed in the preparation of:
  • triazipinones
  • ethyl 2-fluoro-2-benzolyacetate
  • 1,2,4-triazolo[1,5-a]pyrimidine antibiotic essramycin
  • benzo[c]furan-4,7-diones
  • iodonium ylides

Packaging

Packaged in glass bottles

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Asymmetric synthesis of both enantiomers of fluoxetine via microbiological reduction of ethyl benzoylacetate.
Chenevert R, et al.
Tetrahedron, 48(33), 6769-6776 (1992)
Mohamed G Badrey et al.
Molecules (Basel, Switzerland), 17(10), 11538-11553 (2012-09-29)
A number of interesting heterocycles were prepared through interaction of the intermediate 3-amino-8-hydroxy-4-imino-6-methyl-5-phenyl-4,5-dihydro-3H-chromeno-[2,3-d]pyrimidine (1) and reagents such as hydrazonyl halides 2 to furnish triazine derivatives 4a-l. Reaction of 1 with phenacyl bromide afforded compound 5. Moreover, the title compound 1
Ugo Battaglia et al.
Journal of natural products, 73(11), 1938-1939 (2010-09-15)
A short synthesis of the 1,2,4-triazolo[1,5-a]pyrimidine antibiotic essramycin is described involving condensation of aminoguanidine with ethyl benzoylacetate to give an amino-1,2,4-triazole, followed by condensation with ethyl acetoacetate to form the pyrimidone ring.
Kuang-Po Chen et al.
Organic & biomolecular chemistry, 7(19), 4074-4081 (2009-09-19)
A manganese(III)-mediated reaction between 2-benzoyl-1,4-benzoquinones and 1,3-dicarbonyl compounds that produces benzo[c]furan-4,7-diones and anthracene-1,4-diones with high chemoselectivity is described. With ethyl butyrylacetate, by changing the solvent, benzo[c]furan-4,7-diones and anthracene-1,4-diones can be generated in high chemoselectivities. With ethyl benzoylacetate, N ,N-dimethyl acetoacetamide
The reaction of ethyl benzoylacetate with malononitrile: a novel synthesis of some pyridazine, pyridazino [2, 3-a] quinazoline and pyrrole derivatives.
Abdelrazek FM, et al.
Tetrahedron, 57(9), 1813-1817 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service