Skip to Content
Merck
All Photos(2)

Key Documents

C4522

Sigma-Aldrich

Cimetidine

Synonym(s):

SKF-92334

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N6S
CAS Number:
Molecular Weight:
252.34
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

CN\C(NC#N)=N\CCSCc1nc[nH]c1C

InChI

1S/C10H16N6S/c1-8-9(16-7-15-8)5-17-4-3-13-10(12-2)14-6-11/h7H,3-5H2,1-2H3,(H,15,16)(H2,12,13,14)

InChI key

AQIXAKUUQRKLND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

H2 histamine receptor antagonist; I1 imidazoline receptor agonist; anti-ulcer agent. Blocks cancer metastasis by inhibiting the expression of E-selectin on the surface of endothelial cells, thus blocking tumor cell adhesion.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kobayashi et al.
Cancer research, 60(14), 3978-3984 (2000-08-05)
Although the beneficial effect of cimetidine on survival in cancer has been clinically demonstrated in colorectal cancer patients, the mode of action of cimetidine has not been elucidated. In this report, we have demonstrated for the first time that cimetidine
Huan Yang et al.
Molecular medicine (Cambridge, Mass.), 28(1), 57-57 (2022-05-17)
Severe COVID-19 is characterized by pro-inflammatory cytokine release syndrome (cytokine storm) which causes high morbidity and mortality. Recent observational and clinical studies suggest famotidine, a histamine 2 receptor (H2R) antagonist widely used to treat gastroesophageal reflux disease, attenuates the clinical
H van der Goot et al.
European journal of medicinal chemistry, 35(1), 5-20 (2000-03-25)
In this review the histaminergic ligands for the histamine H(1), H(2) and H(3) receptors, which are currently used as tools in pharmacological studies, are described. To study interactions with the histamine H(1) receptor, the H(1) agonist 2-aminoethylthiazole has long since
Sarit Pal et al.
Biology open, 11(7) (2022-07-02)
Histamine exerts its physiological functions through its four receptor subtypes. In this work, we report the subcellular localization of histamine receptor 2 (H2R), a G protein-coupled receptor (GPCR), which is expressed in a wide variety of cell and tissue types.
Hiroki Shimizu et al.
ChemMedChem, 17(16), e202200204-e202200204 (2022-06-14)
We synthesized and experimentally tested the passive permeability of more than thirty tetrapeptides mimicking the N-terminus of the pro-apoptotic protein Smac (Second mitochondria-derived activator of caspases). Each peptide bore one or two unnatural Hydrogen Bond Acceptor-bearing Amino Acid (HBA-AA) residues

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service