Skip to Content
Merck
All Photos(1)

Documents

E14403

Sigma-Aldrich

Ethyl α-bromoisobutyrate

98%

Synonym(s):

Ethyl 2-bromo-2-methylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CBrCOOC2H5
CAS Number:
Molecular Weight:
195.05
Beilstein:
1098947
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.444 (lit.)

bp

65-67 °C/11 mmHg (lit.)

density

1.329 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C)(C)Br

InChI

1S/C6H11BrO2/c1-4-9-5(8)6(2,3)7/h4H2,1-3H3

InChI key

IOLQWGVDEFWYNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl α-bromoisobutyrate is an organic bromoester used as an initiator in controlled radical polymerization reactions.

Application

Atom Transfer Radical Polymerization (ATRP) initiator that will generate a polymer with an ethyl end group.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Niesyto et al.
International journal of molecular sciences, 22(1) (2021-01-06)
In this study, drug nanocarriers were designed using linear copolymers with different contents of cholinium-based ionic liquid units, i.e., [2-(methacryloyloxy)ethyl]trimethylammonium chloride (TMAMA/Cl: 25, 50, and 75 mol%). The amphiphilicity of the copolymers was evaluated on the basis of their critical
Na Wang et al.
Analytical methods : advancing methods and applications, 12(36), 4438-4446 (2020-08-29)
As robust functional polymers, polymer brush-based hybrid nanomaterials have potential application in the highly sensitive determination of tumor markers (TMs). Currently, there are plentiful reports on the polymerization methods of functional polymer brushes. Low ppm ATRP (activators (re)generated by electron
Dongpo Xu et al.
Analytical and bioanalytical chemistry, 412(30), 8371-8378 (2020-10-04)
In this study, a simple and efficient strategy for the construction of hydrangea-like mesoporous WO3 nanoflowers templated using diblock copolymer PS119-PtBA129 was developed. The nanoflower shows good gas sensing properties, especially for 3-hydroxy-2-butanone (3H-2B), which is the signature metabolite of
Hailing He et al.
Materials (Basel, Switzerland), 12(16) (2019-08-10)
The balance of strengthening and toughening of poly (lactic acid) (PLA) has been an intractable challenge of PLA nanocomposite development for many years. In this paper, core-shell nanoparticles consisting of a silica rigid core and poly (butyl acrylate) (PBA) flexible
Wojciech Raj et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-21)
A totally new approach in the synthesis of mixed polymer brushes tethered on polyamide (PA) surfaces is presented herein. As a proof of concept, two types of homopolymers were synthesized in sequential surface-initiated atom transfer radical polymerization (SI-ATRP) reactions: poly(methyl

Articles

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

Protocols

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service