All Photos(1)
About This Item
Linear Formula:
C6H5CH(Br)CO2CH3
CAS Number:
Molecular Weight:
229.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.553 (lit.)
bp
113 °C/3 mmHg (lit.)
density
1.455 g/mL at 25 °C (lit.)
functional group
bromo
ester
phenyl
storage temp.
2-8°C
SMILES string
COC(=O)C(Br)c1ccccc1
InChI
1S/C9H9BrO2/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6,8H,1H3
InChI key
NHFBYYMNJUMVOT-UHFFFAOYSA-N
General description
Methyl α-bromophenylacetate is an ester. Palladium catalyzed homocoupling reaction of methyl α-bromophenylacetate with 3,5-dimethyl phenyl boronic acid is reported.
Application
Methyl α-bromophenylacetate may be used as initiator during ESR study of atom transfer radical polymerization (ATRP) of methyl methacrylate (MMA) and ethylene glycol dimethacrylate (EGDMA). It may be used as reagent in the homopolymerization of MMA in supercritical carbon dioxide.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
ESR study on diffusion-controlled atom transfer radical polymerization of methyl methacrylate and ethylene glycol dimethacrylate.
Wang AR and Zhu S.
Macromolecules, 35(27), 9926-9933 (2002)
A novel palladium-catalyzed homocoupling reaction initiated by transmetallation of palladium enolates.
Lei A and Zhang X.
Tetrahedron Letters, 43(14), 2525-2528 (2002)
Atom transfer radical polymerization of MMA with a macromolecular ligand in a fluorinated solvent and in supercritical carbon dioxide.
Grignard B, et al.
Eur. Polymer J., 44(3), 861-871 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service