Skip to Content
Merck
All Photos(2)

Key Documents

482250

Sigma-Aldrich

4,4′-Dinonyl-2,2′-dipyridyl

97%

Synonym(s):

dNbpy

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44N2
CAS Number:
Molecular Weight:
408.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

61-63 °C (lit.)

SMILES string

CCCCCCCCCc1ccnc(c1)-c2cc(CCCCCCCCC)ccn2

InChI

1S/C28H44N2/c1-3-5-7-9-11-13-15-17-25-19-21-29-27(23-25)28-24-26(20-22-30-28)18-16-14-12-10-8-6-4-2/h19-24H,3-18H2,1-2H3

InChI key

VHJFWJXYEWHCGD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maryam Bozorg et al.
Soft matter, 16(4), 1066-1081 (2019-12-21)
Poly[oligo(ethylene oxide)] based gradient and random copolymers with different compositions are synthesized via Cu-based atom transfer radical polymerization. The solubility behavior of these copolymers in pure water and in the presence of different salts, surfactants and ethanol is investigated. According
Thien Van Truong et al.
Scientific reports, 10(1), 12199-12199 (2020-07-24)
A new technique for promoting nucleation and growth of membrane protein (MP) crystals from micellar environments is reported. It relies on the conjugation of micelles that sequester MPs in protein detergent complexes (PDCs). Conjugation via amphiphilic [metal:chelator] complexes presumably takes
Tomy Muringayil Joseph et al.
Polymers, 12(1) (2020-01-18)
The copolymerization of styrene (St) with a bioderived monomer, pentadecylphenyl methacrylate (PDPMA), via atom transfer radical polymerization (ATRP) was studied in this work. The copolymerization reactivity ratio was calculated using the composition data obtained from 1H NMR spectroscopy, applying Kelen-Tudos
Yi-Shen Huang et al.
Polymers, 11(6) (2019-06-27)
Atom transfer radical polyaddition (ATRPA) was utilized herein to synthesize a specific functional polyester. We conducted ATRPA of 4-vinylbenzyl 2-bromo-2-phenylacetate (VBBPA) inimer and successfully obtained a linear type poly(VBBPA) (PVBBPA) polyester with benzylic bromides along the backbone. To obtain a
Wojciech Raj et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-21)
A totally new approach in the synthesis of mixed polymer brushes tethered on polyamide (PA) surfaces is presented herein. As a proof of concept, two types of homopolymers were synthesized in sequential surface-initiated atom transfer radical polymerization (SI-ATRP) reactions: poly(methyl

Articles

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

See All

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service