Skip to Content
Merck
All Photos(2)

Key Documents

92260

Sigma-Aldrich

Trimethylamine solution

31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer

Synonym(s):

N,N,N-Trimethylamine, N,N-Dimethylmethanamine, TMA solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N
CAS Number:
Molecular Weight:
59.11
Beilstein:
956566
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

Quality Level

contains

toluene as stabilizer

concentration

31-35 wt. % in ethanol
4.2 M

functional group

amine

SMILES string

CN(C)C

InChI

1S/C3H9N/c1-4(2)3/h1-3H3

InChI key

GETQZCLCWQTVFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethylamine ethanolic solution is used in the preparation of:
  • Glycidyltrimethylammonium bromide.[1]
  • Trimethylammonium salt of ethylenediaminetetraacetic acid (EDTA).[2]
  • Quaternized ammonium chlorides that exhibit antimicrobial activity.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Glycidyltrimethylammonium chloride and related compounds.
McClure JD.
The Journal of Organic Chemistry, 35(6), 2059-2061 (1970)
Preparation and antimicrobial behaviour of quaternary ammonium thiol derivatives able to be grafted on metal surfaces.
Thebault P, et al.
European Journal of Medicinal Chemistry, 44(2), 717-724 (2009)
Demineralization in organic solvents by alkylammonium salts of ethylenediaminetetra-acetic acid.
Scott JE and Kyffin TW
The Biochemical Journal, 169(3), 697-701 (1978)
S L Hillier
American journal of obstetrics and gynecology, 169(2 Pt 2), 455-459 (1993-08-01)
Bacterial vaginosis is characterized microbiologically by replacement of the Lactobacillus-predominant vaginal flora by Gardnerella vaginalis, Bacteroides species, Mobiluncus species, and genital mycoplasmas. A standardized, laboratory-based diagnostic test for bacterial vaginosis is desirable in those instances in which a microscope is
M Al-Waiz et al.
Clinical pharmacology and therapeutics, 42(5), 588-594 (1987-11-01)
Trimethylamine (TMA) and its N-oxide (TMAO) are normal components of human urine. They are present in the diet and also derived from the enterobacterial metabolism of precursors such as choline. Dietary TMA is almost entirely metabolized to and excreted as

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service