Skip to Content
Merck
All Photos(1)

Key Documents

243205

Sigma-Aldrich

Trimethylamine

anhydrous, ≥99%

Synonym(s):

N,N,N-Trimethylamine, N,N-Dimethylmethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N
CAS Number:
Molecular Weight:
59.11
Beilstein:
956566
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

2.09 (vs air)

vapor pressure

13.3 psi ( 21 °C)

Assay

≥99%

autoignition temp.

374 °F

expl. lim.

11.6 %

impurities

≤0.05% ammonia
≤0.05% methylamine
≤0.2% dimethylamine

bp

3-4 °C (lit.)

mp

−117 °C (lit.)

density

0.63 g/mL at 20 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CN(C)C

InChI

1S/C3H9N/c1-4(2)3/h1-3H3

InChI key

GETQZCLCWQTVFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethylamine (TMA) has been used in the preparation of poly[9,9′-bis(6′-N,N,N-trimethylammonium)hexyl)fluorene-co-alt-4,7-(2,1,3-benzothiadiazole) dibromide] (PFBT), a water-soluble, cationic conjugated polymer used in label-free DNA microarrays.[1] It can also be used to prepare benzyltrimethylammonium chloride, which then reacts with sodium ethoxide to form benzyltrimethylammonium ethoxide.[2] The adsorption of TMA on the gold surface of trimethylsilylated barium nitrate-gold/titanosilicate catalyst acts as a promoter for the propylene epoxidation with oxygen and hydrogen.[3]

Packaging

Supplied in a Sure/Pac cylinder and has a 316 stainless steel needle valve with a male 1/4" NPTF outlet thread installed . Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trimethylamine as a Gas?Phase Promoter: Highly Efficient Epoxidation of Propylene over Supported Gold Catalysts.
Chowdhury B, et al.
Angewandte Chemie (International Edition in English), 45(3), 412-415 (2006)
Benzyltrimethylammonium Ethoxide: Ammonium, benzyltrimethyl?, ethoxide
Croxall W J, et al.
Organic Syntheses, 38, 5-5 (2003)
Bin Liu et al.
Nature protocols, 1(4), 1698-1702 (2007-05-10)
We describe the synthesis of poly[9,9'-bis(6''-N,N,N-trimethylammonium)hexyl)fluorene-co-alt-4,7-(2,1,3-benzothiadiazole) dibromide] (PFBT), a cationic, water-soluble conjugated polymer used in label-free DNA microarrays. This polymer was designed to have a maximum absorbance of close to 488 nm, which meets the excitation wavelength of most commercial
M Al-Waiz et al.
Clinical pharmacology and therapeutics, 42(5), 588-594 (1987-11-01)
Trimethylamine (TMA) and its N-oxide (TMAO) are normal components of human urine. They are present in the diet and also derived from the enterobacterial metabolism of precursors such as choline. Dietary TMA is almost entirely metabolized to and excreted as
S L Hillier
American journal of obstetrics and gynecology, 169(2 Pt 2), 455-459 (1993-08-01)
Bacterial vaginosis is characterized microbiologically by replacement of the Lactobacillus-predominant vaginal flora by Gardnerella vaginalis, Bacteroides species, Mobiluncus species, and genital mycoplasmas. A standardized, laboratory-based diagnostic test for bacterial vaginosis is desirable in those instances in which a microscope is

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service