Skip to Content
Merck
All Photos(2)

Key Documents

426458

Sigma-Aldrich

Dimethylamine solution

40 wt. % in H2O

Synonym(s):

DMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH
CAS Number:
Molecular Weight:
45.08
Beilstein:
605257
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.55 (vs air)

vapor pressure

16.97 psi ( 55 °C)
212 mmHg ( 20 °C)

form

liquid

autoignition temp.

753 °F

concentration

38-42% (with HCl, titration)
40 wt. % in H2O

refractive index

n20/D 1.37

density

0.89 g/mL at 25 °C

functional group

amine

SMILES string

CNC

InChI

1S/C2H7N/c1-3-2/h3H,1-2H3

InChI key

ROSDSFDQCJNGOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethylamine is utilized as a building block or reagent in the production of polyurethanes, ion-exchange resins, and dyes.

Application

Dimethylamine (DMA) solution can be used:
  • To synthesize Ge7O14.5F2‚[(CH3)2NH2]3(H2O)0.86, a germanate-based porous inclusion complex.
  • To synthesize ethyl 2-dimethylamino-4,6-diarylpyrimidine-5-carboxylates.
  • To convert 5′-bromo-2,2′-bithiophene-5carboxaldehyde to 5′-dimethylamino-2,2′-bithiophene-5-carboxaldehyde.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Porous Germanates: Synthesis, Structure, and Inclusion Properties of Ge7O14.5F2.[(CH3)2NH2]3(H2O) 0.86.
Li H, et al.
Journal of the American Chemical Society, 120(33), 8567-8568 (1998)
Polymer-assisted synthesis of ethyl 2-amino-4, 6-diarylpyrimidine-5-carboxylates.
Eynde V, et al.
ARKIVOC (Gainesville, FL, United States), 15, 2003-2003 (2003)
Ping Yan et al.
The Journal of organic chemistry, 73(17), 6587-6594 (2008-07-31)
There is a growing need for cellular imaging with fluorescent probes that emit at longer wavelengths to minimize the effects of absorption, autofluorescence, and scattering from biological tissue. In this paper a series of new environmentally sensitive hemicyanine dyes featuring
Effective recovery of Au from low-concentration solutions by a self-synthesized mesoporous resin modified by dimethylamine
Xu W, et al.
Industrial & Engineering Chemistry Research, 61(7), 2894-2903 (2021)
C Mulder et al.
Journal of neural transmission (Vienna, Austria : 1996), 109(9), 1203-1208 (2002-08-31)
Nitric oxide (NO) may play a role in the pathophysiology of Alzheimer's disease (AD). Asymmetric dimethylarginine (ADMA), an endogenous inhibitor of NO synthase, is involved in regulation of NO production. Recently it has been reported that dimethylarginine dimethylaminohydrolase, an enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service