Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trimethylamine solution
Linear Formula:
(CH
3
)
3
N
CAS Number:
75-50-3
Molecular Weight:
59.11
Products
Product Category
aroma chemicals (4)
building blocks (2)
Brand
Sigma-Aldrich (6)
Biological Source
Formula Weight
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Kosher (3)
Food Grade (2)
Purum (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
92262
Trimethylamine solution,
43.0-49.0% in H
2
O (T)
Expand
View Pricing
92260
Trimethylamine solution,
31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Expand
View Pricing
W324108
Trimethylamine solution,
25 wt. % in H
2
O
Expand
View Pricing
W324125
Trimethylamine solution,
25 wt. % in H
2
O, FG
Expand
View Pricing
W324110
Trimethylamine solution,
45 wt. % in H
2
O, FG
Expand
View Pricing
W550132
Trimethylamine solution,
25 wt. % in propylene glycol
Expand
View Pricing