Skip to Content
Merck
All Photos(1)

Key Documents

860646

Sigma-Aldrich

1-Methyl-DL-tryptophan

97%

Synonym(s):

1-Methyl-DL-tryptophan, 1-Methyltryptophan, 2-Amino-3-(1-methyl-1H-indol-3-yl)propanoic acid, 2-Amino-3-(1-methyl-1H-indol-3-yl)propanoicacid, 2-Amino-3-(1-methylindol-3-yl)propanoic acid, DL-1-Methyltryptophan, N′-Methyl-DL-tryptophan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

250 °C (dec.) (lit.)

SMILES string

Cn1cc(CC(N)C(O)=O)c2ccccc12

InChI

1S/C12H14N2O2/c1-14-7-8(6-10(13)12(15)16)9-4-2-3-5-11(9)14/h2-5,7,10H,6,13H2,1H3,(H,15,16)

InChI key

ZADWXFSZEAPBJS-UHFFFAOYSA-N

Application

1-Methyl-DL-tryptophan, a competitive inhibitor of indoleamine 2,3 dioxygenase-1 (IDO1), has been used to study inflammation-induced depression in mice and syncytial virus induced bronchiolitis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Böttcher et al.
Stem cells (Dayton, Ohio), 34(2), 516-521 (2015-10-21)
Mesenchymal stromal cells (MSCs) possess numerous regenerative and immune modulating functions. Transplantation across histocompatibility barriers is feasible due to their hypo-immunogenicity. MSCs have emerged as promising tools for treating graft-versus-host disease following allogeneic stem cell transplantation. It is well established
Qi-Xiang Ye et al.
Experimental and therapeutic medicine, 14(3), 1884-1891 (2017-10-01)
Cytotoxic T-lymphocyte-associated protein 4 immunoglobulin (CTLA4Ig) and anti-cluster of differentiation 154 (anti-CD154) are able to block B7/CD28 and CD40/CD154 co-stimulatory signals in T cells. Additionally, they promote hematopoietic stem cell transplantation (HSCT) in sensitized recipients and are able to induce
Seung-Ha Yang et al.
Experimental & molecular medicine, 41(5), 315-324 (2009-03-25)
Mesenchymal stem cells (MSCs) can inhibit T cell proliferation; however, the underlying mechanisms are not clear. In this study, we investigated the mechanisms of the immunoregulatory activity of MSCs on T cells. Irradiated MSCs co-cultured with either na?ve or pre-activated
Motivational changes that develop in a mouse model of inflammation-induced depression are independent of indoleamine 2, 3 dioxygenase.
Vichaya, Elisabeth G et al.
Neuropsychopharmacology (2018)
Neda Milosavljevic et al.
Liver transplantation : official publication of the American Association for the Study of Liver Diseases and the International Liver Transplantation Society, 23(8), 1040-1050 (2017-05-10)
Mesenchymal stem cells (MSCs) are, due to immunomodulatory characteristics, considered as novel agents in the treatment of immune-mediated acute liver failure. Although it is known that MSCs can regulate activation of T lymphocytes, their capacity to modulate function of neutrophils

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service