Skip to Content
Merck
All Photos(1)

Key Documents

G11950

Sigma-Aldrich

Guanine

98%

Synonym(s):

2-Amino-1,7-dihydro-6H-purin-6-one, 2-Amino-6-hydroxypurine, 2-Aminohypoxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5O
CAS Number:
Molecular Weight:
151.13
Beilstein:
9680
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

>300 °C (lit.)

solubility

hydrochloric acid: soluble 5 M, clear to slightly hazy, colorless to faintly yellow-green

SMILES string

NC1=Nc2[nH]cnc2C(=O)N1

InChI

1S/C5H5N5O/c6-5-9-3-2(4(11)10-5)7-1-8-3/h1H,(H4,6,7,8,9,10,11)

InChI key

UYTPUPDQBNUYGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adenine, cytosine, thymine and guanine are the four main nucleobases found in nucleic acids DNA and RNA. Guanine is a purine derivative. It is reported to assemble into square-planar groups that resemble macrocycles, in which the bases interact via hydrogen bonds. In DNA and RNA, stretches of guanine bases are reported to form stable four stranded helices in the presence of sodium or potassium ions. The electrochemical oxidation of guanine has been studied in aqueous media at various carbon electrodes. It reacts with peroxynitrite under physiological conditions to afford 8-nitroguanine.

Application

Guanine is suitable reagent used to investigate the mechanism of electrochemical oxidation of guanine and adenine using a glassy carbon microelectrode and cyclic and differential pulse voltammetry. It may be used in the preparation of mixed-sequence peptide nucleic acid (PNA) monomers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Oliveira-Brett et al.
Bioelectrochemistry (Amsterdam, Netherlands), 55(1-2), 61-62 (2002-01-12)
The electrochemical oxidation mechanism of guanine and adenine was investigated using a glassy carbon microelectrode and cyclic and differential pulse voltammetry. It is pH-dependent and the electron transfer process occurs in consecutive steps with the formation of strongly adsorbed dimers
K Phillips et al.
Journal of molecular biology, 273(1), 171-182 (1997-11-21)
In both DNA and RNA, stretches of guanine bases can form stable four-stranded helices in the presence of sodium or potassium ions. Sequences with a propensity to form guanine tetraplexes have been found in chromosomal telomers, immunoglobulin switch regions, and
Qian Li et al.
The journal of physical chemistry. B, 114(21), 7423-7428 (2010-05-08)
The electrochemical oxidation of guanine is studied in aqueous media at various carbon electrodes. Specifically edge plane pyrolytic graphite (EPPG), basal plane pyrolytic graphite (BPPG), and highly ordered pyrolytic graphite (HOPG) were used, and the voltammetry was found to vary
V Yermilov et al.
Carcinogenesis, 16(9), 2045-2050 (1995-09-01)
Nitric oxide and superoxide anion, both formed in inflamed tissues, react rapidly to form the peroxynitrite anion (ONOO-), a strong oxidant which can initiate reactions characteristic of hydroxyl radical (HO.), nitronium ion (NO2+) and nitrogen dioxide radical (NO2.). Peroxynitrite, therefore
Synthesis of peptide nucleic acid monomers containing the four natural nucleobases: thymine, cytosine, adenine, and guanine and their oligomerization.
Dueholm KL, et al.
The Journal of Organic Chemistry, 59(19), 5767-5773 (1994)

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service