Skip to Content
Merck
All Photos(4)

Key Documents

C3506

Sigma-Aldrich

Cytosine

≥99%

Synonym(s):

4-Amino-2-hydroxypyrimidine, 4-Aminopyrimidin-2-(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3O
CAS Number:
Molecular Weight:
111.10
Beilstein:
2637
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥99%

form

powder

mp

>300 °C (lit.)

solubility

0.5 M HCl: 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

SMILES string

NC1=NC(=O)NC=C1

InChI

1S/C4H5N3O/c5-3-1-2-6-4(8)7-3/h1-2H,(H3,5,6,7,8)

InChI key

OPTASPLRGRRNAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cytosine has been used:
  • for the preparation of nucleobase solutions
  • as a standard for high-performance liquid chromatography (HPLC)
  • for the estimation of global methylation rate
  • for nucleoside 5′-triphosphate (NTP) synthesis
  • purification

Biochem/physiol Actions

Cytosine (C) is one of the four main bases found in DNA and RNA, along with adenine, guanine, and thymine (uracil in RNA).
Cytosine is a pyrimidine, which forms three hydrogen bonds to base pair with guanine. It forms a nucleotide cytidine, that is phosphorylated to cytidine 5′ monophosphate (CMP), cytidine 5′ diphosphate (CDP) and cytidine 5′ triphosphate (CTP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deuterated nucleotides as chemical probes of RNA structure: a detailed protocol for the enzymatic synthesis of a complete set of nucleotides specifically deuterated at ribose carbons
Azad R, et al.
ScienceOpen Research (2015)
Encyclopedia of Genetics (2001)
Nucleobase sensing using highly-sensitive surface-enhanced Raman spectroscopy templates comprising organic semiconductor peptide nanotubes and metal nanoparticles
Almohammed S, et al.
Sensing and Bio-Sensing Research, 24, 100287-100287 (2019)
Akira Ono et al.
Chemical Society reviews, 40(12), 5855-5866 (2011-08-10)
Pyrimidine base pairs in DNA duplexes selectively capture metal ions to form metal ion-mediated base pairs, which can be evaluated by thermal denaturation, isothermal titration calorimetry, and nuclear magnetic resonance spectroscopy. In this critical review, we discuss the metal ion
Irene M Kaplow et al.
Genome research, 25(6), 907-917 (2015-04-26)
DNA methylation is an epigenetic modification that plays a key role in gene regulation. Previous studies have investigated its genetic basis by mapping genetic variants that are associated with DNA methylation at specific sites, but these have been limited to

Articles

A shortened review of fundamentals of Porous Graphitic Carbon (PGC) materials. What they are and how do they work as a stationary phase material in HPLC?.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service