Skip to Content
Merck
All Photos(4)

Key Documents

283223

Sigma-Aldrich

Poly(allylamine hydrochloride)

average Mw 50,000

Synonym(s):

PAA HCl, PAH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CH2NH2 · HCl)]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

mol wt

average Mw 50,000

SMILES string

Cl.NCC=C

InChI

1S/C3H7N/c1-2-3-4/h2H,1,3-4H2

InChI key

VVJKKWFAADXIJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to make redox hydrogel-modified electrodes for measuring enzyme responses.

Packaging

The polyelectrolyte poly(allylamine hydrochloride) has been used in the preparation of an antireflective coating consisting of alternating layers of poly (allyamine hydrochloride) and silica nanoparticles

Other Notes

May contain up to ≤10% residual monomer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yancey, S.E. et al
Journal of Applied Physics, 99, 034313-034313 (2006)
Analytical Chemistry, 68, 4186-4186 (1990)
Magdalena Oćwieja et al.
Langmuir : the ACS journal of surfaces and colloids, 29(11), 3546-3555 (2013-01-17)
Systematic studies of silver particle deposition kinetics under diffusion transport on poly(allylamine hydrochloride) (PAH) modified mica were carried out. Monolayer coverage, quantitatively determined by AFM and SEM, was regulated within broad limits by adjusting the deposition time and the ionic
Ya Wang et al.
Macromolecular bioscience, 12(10), 1321-1325 (2012-09-12)
Fluorescent-magnetic-biotargeting multifunctional microcapsules (FMBMMs) are designed and fabricated via layer-by-layer assembly. It is found that the arginine-glycine-aspartate-modified FMBMMs were capable of sensitively detecting and efficiently isolating approximately 80% target cancer cells within 20 min. More importantly, FMBMMs present a general
Zhi-Lu Yang et al.
Journal of biomedical materials research. Part A, 100(11), 3124-3133 (2012-07-21)
A simple method is developed to construct anticoagulant surfaces via passive adsorption of heparin onto the protonated plasma-polymerized allylamine (PPAam) films from phosphate-buffered saline (PBS). These protonated PPAam surfaces are found to have high affinity to heparin. Importantly, the heparin-functionalized

Articles

Recently, layer-by-layer (LbL) assembly has emerged as a versatile, gentle and, simple method for immobilization of functional molecules in an easily controllable thin film morphology.1,2 In this short review, we introduce recent advances in functional systems fabricated by using the mild, yet adaptable LbL technique.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service