Skip to Content
Merck
All Photos(1)

Key Documents

701491

Sigma-Aldrich

Silicon dioxide, alumina doped

nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, ≥99.9% trace metals basis

Synonym(s):

silicic anhydride, alumina doped preparation

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(SiO2)x(Al2O3)y
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9% trace metals basis

form

dispersion
nanoparticles

composition

aluminum, 7.5 wt. %

concentration

20 wt. % in H2O

surface area

75.5 m2/g , BET

particle size

<50 nm

pH

4-6(neat)

density

1.087 g/mL at 25 °C

SMILES string

O=[Si]=O.O=[Al]O[Al]=O

InChI

1S/2Al.O2Si.3O/c;;1-3-2;;;

InChI key

HNPSIPDUKPIQMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Contains 0.025 wt. % Ammonium hydroxide.

Legal Information

Product of Engi-Mat Co.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Yeong Won et al.
Journal of biomedical nanotechnology, 9(5), 886-890 (2013-06-28)
In this study, we developed a simple and fast isolation tool of Escherichia coli O157:H7 (E. coli O157:H7) using a magnet nanoparticle embedded silica nanotube (MNSNT) for the detection of E. coli O157:H7 in the sample with nucleic acid based
David M Brown et al.
Toxicology letters, 224(1), 147-156 (2014-01-28)
We investigated the effects of silica particles and nanoparticles (NPs) (50 nm and 200 nm) with a neutral and positively charged surface when dispersed in saline, bovine serum albumin (BSA) or lung lining fluid (LLF) 24 h post instillation into
Jeong Ha Yoo et al.
Journal of nanoscience and nanotechnology, 13(11), 7615-7619 (2013-11-20)
Recently, magnetic and luminescent composite silica with structure of micro- and nanospheres containing both magnetic (Fe3O4) nanoparticles (MPs) and quantum dots (QDs) has attracted great interests. In this study, we have prepared core-shell structure of silica spheres in which magnets
K E Kovalenko et al.
Ecological applications : a publication of the Ecological Society of America, 23(5), 1048-1060 (2013-08-24)
Boreal wetlands play an important role in global carbon balance. However, their ecosystem function is threatened by direct anthropogenic disturbance and climate change. Oil sands surface mining in the boreal regions of Western Canada denudes tracts of land of organic
Kyle Steenland et al.
CA: a cancer journal for clinicians, 64(1), 63-69 (2013-12-12)
Silica has been known to cause silicosis for centuries, and evidence that silica causes lung cancer has accumulated over the last several decades. This article highlights 3 important developments in understanding the health effects of silica and preventing illness and

Articles

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Recent demand for electric and hybrid vehicles, coupled with a reduction in prices, has caused lithium-ion batteries (LIBs) to become an increasingly popular form of rechargeable battery technology.

Silica's versatility spans various industries, including biomedical applications.

Silica's versatility spans various industries, including biomedical applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service