Skip to Content
Merck
All Photos(2)

Key Documents

643653

Sigma-Aldrich

Aluminosilicate, mesostructured

MCM-41 (hexagonal)

Synonym(s):

MCM-41 molecular sieves

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(SiO2)x(Al2O3)y
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

aluminum, ~3%

surface area

900-1100 m2/g , BET

unit cell size

4.5-4.8 nm

pore size

0.8-1.1 cm3/g pore volume
2-4 nm pore size

application(s)

battery manufacturing

SMILES string

O=[Si]=O.O=[Al]O[Al]=O

InChI

1S/2Al.O2Si.3O/c;;1-3-2;;;

InChI key

HNPSIPDUKPIQMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aluminosilicate, mesostructured (Al-MSU) is an analog of hexagonal MCM-41 that can be assembled by zeolite seeds. It is hydrothermally stable and strongly acidic, which makes it useful in catalysis and adsorption technology.

Application

Al-MSU can be used as an adsorbent in the formation of hybrid membranes for fuel cell applications. It can also be used as a catalyst for biomass pyrolysis and production of aromatic amine antioxidants.

Formula variant

(SiO2)0.9875(Al2O3)0.0125­­·xH2O

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aluminosilicate mesostructures with improved acidity and hydrothermal stability
Liu Y and Pinnavaia TJ
Journal of Materials Chemistry, 12(11), 3179-3190 (2002)
Mesostructured aluminosilicate alkylation catalysts for the production of aromatic amine antioxidants
Liu Y, et al.
J. Catal., 225(2), 381-387 (2004)
Steam-stable aluminosilicate mesostructures assembled from zeolite type Y seeds
Liu Y, et al.
Journal of the American Chemical Society, 122(36), 8791-8792 (2000)
Hydrothermally stable mesoporous aluminosilicates (MSU-S) assembled from zeolite seeds as catalysts for biomass pyrolysis
Triantafyllidis KS, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 99(1-2), 132-139 (2007)
Mesostructured-aluminosilicate-Nafion hybrid membranes for direct methanol fuel cells
Meenakshi S, et al.
Electrochimica Acta, 89(11), 35-44 (2013)

Articles

Mesoporous Materials include a range of high surface area porous silicates with applications in gas adsorption, drug delivery, diagnostics and catalysis.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service