Skip to Content
Merck
All Photos(2)

Key Documents

S5505

Sigma-Aldrich

Silica, fumed

powder, 0.2-0.3 μm avg. part. size (aggregate)

Synonym(s):

Silica, Silicic anhydride, Silicon dioxide, Silicon dioxide amorphous

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

powder

surface area

200 m2/g (±25 m2/g)

refractive index

n20/D 1.46 (lit.)

avg. part. size

0.2-0.3 μm (aggregate)

density

2.3 lb/cu.ft at 25 °C (bulk density)(lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fumed Silica (SiO2) has been used as a precursor to synthesize:
  • Chlorosilanes via silanization, which are used to prepare silica/polydimethylsiloxane membranes to study permeation performance.
  • Silicon carbide (SiC) nanoparticles by sol-gel method.
  • Calcium silicate hydrate (C-S-H) by treating with CaO and water via hydrothermal reaction.
  • Si/PtO2 catalyst for the selective conversion of cinnamaldehyde to cinnamic acid by aerobic oxidation.


It can be also used in the preparation of:
  • Silica sulfuric acid catalyst used in the synthesis of 3,4-dihydropyrimidin-2(1H)-ones via one-pot three-component Biginelli reaction.
  • HClO4-SiO2 heterogeneous catalyst used in the synthesis of tetrasubstituted imidazoles.

Features and Benefits

Has interesting thickening and thixotropic properties, and an enormous external surface area.

Other Notes

Particle sizes are for primary particles, which form branched, chain-like aggregates a few tenths of a micron long.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Organosilane modified silica/polydimethylsiloxane mixed matrix membranes for enhanced propylene/nitrogen separation
Beltran AB, et al.
Applied Surface Science, 258(1), 337-345 (2011)
Synthesis of nano-structured 3C-SiC by carbothermal reduction of silicon bearing gel and carbon soot
Sharma M, et al.
Materials Letters, 65(14), 2161-2164 (2011)
Platinum catalysed aerobic selective oxidation of cinnamaldehyde to cinnamic acid
Durndell LJ, et al.
Catalysis Today, 333(4), 161-168 (2019)
Silica sulfuric acid: an efficient and reusable catalyst for the one-pot synthesis of 3, 4-dihydropyrimidin-2 (1H)-ones
Salehi P, et al.
Tetrahedron Letters, 44(14), 2889-2891 (2003)
Effect of C/S ratio on morphology and structure of hydrothermally synthesized calcium silicate hydrate
He Y, et al.
Journal of Wuhan University of Technology-Materials Science Edition, 26(4), 770-773 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service