Skip to Content
Merck
All Photos(3)

Key Documents

637238

Sigma-Aldrich

Silicon dioxide

nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis

Synonym(s):

Silica, Nanosilica, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5% trace metals basis

form

nanopowder

particle size

10-20 nm (BET)

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

density

2.2-2.6 g/mL at 25 °C

bulk density

0.011 g/mL

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The refractive index of silica nanoparticles is similar to polymeric coatings, so it finds several applications in the preparations of nanocomposites. Properties suh as, thermal resistance, electrical and mechanical are enhanced. Silica nanoparticles find uses in various fields of catalysts, pigment stabilization, electronics and sensors.

Application

Silica nanoparticles have been used :
  • To study the nonlinear optical properties of SiO2 nanocomposites in weak optical fields.
  • As a nanoadditive in nanocomposite calcium alginate fibers.
  • As dispersant in liquid polymethylsiloxane to exihibit unique optical non linearity in low intensity visible radiation fields.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

APS-silane modification of silica nanoparticles: effect of treatm,ent's variables on the grafting content and colloidal stability of nanoparticles
Journal of Coatings Technology and Research, 11(4), 651-660 (2014)
Unusual nonlinear optical properties of SiO2 nanocomposite in weak optical fields
Milichko VA, et al.
Applied Physics. A, Materials Science & Processing, 111(1), 319-322 (2013)
Nanocomposite calcium alginate fibres containing SiO2 and bioglass
Bogun M, et al.
Spine, 12, 18-18 (2010)
Saisai Wei et al.
The international journal of biochemistry & cell biology, 113, 95-102 (2019-06-21)
ZnO nanoparticle (ZnO NP) exposure causes oxidative stress in the respiratory system, leading to pulmonary damage. Activating transcription factor 3 (ATF3) participates in a variety of cellular stress responses. However, the role of ATF3 in ZnO NP genotoxicity and cytotoxicity
David M Brown et al.
Toxicology letters, 224(1), 147-156 (2014-01-28)
We investigated the effects of silica particles and nanoparticles (NPs) (50 nm and 200 nm) with a neutral and positively charged surface when dispersed in saline, bovine serum albumin (BSA) or lung lining fluid (LLF) 24 h post instillation into

Articles

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Recent demand for electric and hybrid vehicles, coupled with a reduction in prices, has caused lithium-ion batteries (LIBs) to become an increasingly popular form of rechargeable battery technology.

Silica's versatility spans various industries, including biomedical applications.

Silica's versatility spans various industries, including biomedical applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service