Skip to Content
Merck
All Photos(1)

Key Documents

647101

Sigma-Aldrich

Silicon

wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm

Synonym(s):

Silicon element

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si
CAS Number:
Molecular Weight:
28.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

crystalline (cubic (a = 5.4037))
wafer (single side polished)

Quality Level

does not contain

dopant

diam. × thickness

2 in. × 0.5 mm

bp

2355 °C (lit.)

mp

1410 °C (lit.)

density

2.33 g/mL at 25 °C (lit.)

semiconductor properties

<111>, N-type

SMILES string

[Si]

InChI

1S/Si

InChI key

XUIMIQQOPSSXEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Silicon Photonics and Electronics: Discusses the integration of silicon photonics with electronics, relevant for developing high-performance computational devices, pertinent to material science and high-performance computing research (C Xiang et al., 2021).

  • Silicon Silicon pi Single Bond: Details the structural chemistry of silicon and its implications in molecular and material sciences, useful for chemists interested in silicon′s applications in nanotechnology and materials science (S Kyushin et al., 2020).

Physical properties

0 vortex defects. Etch pitch density (EPD) < 100 (cm-2). Resistivity 100 - 3000 Ωcm.
Oxygen content: ≤ 1~1.8 x 1018 /cm3; Carbon content: ≤ 5 x 1016 /cm3; Boule diameter: 1~8 ″

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariya Nazish Memon et al.
JPMA. The Journal of the Pakistan Medical Association, 62(12), 1329-1332 (2013-07-23)
To evaluate the outcome of nasolacrimal intubation as a primary treatment of congenital nasolacrimal duct obstruction (NLDO) in children up to 4 years of age. During the 3 years period from July 2008 to June 2011, in the Paediatric Ophthalmology
Min Joon Huang et al.
Journal of nanoscience and nanotechnology, 13(6), 3810-3817 (2013-07-19)
In this work, we demonstrated a silicon nanowire (SiNW) biosensing platform capable of simultaneously identifying different Dengue serotypes on a single sensing chip. Four peptide nucleic acids (PNAs), specific to each Dengue serotypes (DENV-1 to DENV-4), were spotted on different
Bo-Soon Kim et al.
Journal of nanoscience and nanotechnology, 13(5), 3622-3626 (2013-07-19)
A subwavelength structure (SWS) was formed via a simple chemical wet etching using a gold (Au) catalyst. Single nano-sized Au particles were fabricated by metallic self-aggregation. The deposition and thermal annealing of the thin metallic film were carried out. Thermal
Jaewoo Lee et al.
Journal of nanoscience and nanotechnology, 13(5), 3495-3499 (2013-07-19)
A spin-casting process for fabricating polycrystalline silicon sheets for use as solar cell wafers is proposed, and the parameters that control the sheet thickness are investigated. A numerical study of the fluidity of molten silicon indicates that the formation of
Seungil Park et al.
Journal of nanoscience and nanotechnology, 13(5), 3397-3402 (2013-07-19)
We investigated the thin film growths of hydrogenated silicon by hot-wire chemical vapor deposition with different flow rates of SiH4 and H2 mixture ambient and fabricated thin film solar cells by implementing the intrinsic layers to SiC/Si heterojunction p-i-n structures.

Articles

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

See All

Protocols

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service