Skip to Content
Merck
All Photos(1)

Key Documents

646687

Sigma-Aldrich

Silicon

wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm

Synonym(s):

Silicon element

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si
CAS Number:
Molecular Weight:
28.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

crystalline (cubic (a = 5.4037))
wafer (single side polished)

does not contain

dopant

diam. × thickness

2 in. × 0.5 mm

bp

2355 °C (lit.)

mp

1410 °C (lit.)

density

2.33 g/mL at 25 °C (lit.)

semiconductor properties

<100>, N-type

SMILES string

[Si]

InChI

1S/Si

InChI key

XUIMIQQOPSSXEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silicon wafers or a “slice” of substrate find applications in the fabrication of integrated circuits, solar cells etc. They serve as a substrate for various microelectronic devices.

Application

  • Innovative Solutions for High-Performance Silicon Anodes in Lithium-Ion Batteries: Overcoming Challenges and Real-World Applications.: This article addresses the challenges and real-world applications of high-performance silicon anodes in lithium-ion batteries, presenting innovative solutions to enhance their efficiency (Khan et al., 2024).
  • Strain Engineering: Perfecting Freestanding Perovskite Oxide Fabrication.: This research focuses on strain engineering to improve the fabrication of freestanding perovskite oxides, which are crucial for various high-purity silicon applications in academia (Yun et al., 2024).

Physical properties

0 vortex defects. Etch pitch density (EPD) < 100 (cm-2). Resistivity 100 - 3000 Ωcm
Oxygen content: ≤ 1~1.8 x 1018 /cm3; Carbon content: ≤ 5 x 1016 /cm3; Boule diameter: 1~8 ″

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular monolayers on silicon surfaces.
Lopinski GP and Wayner DDM.
Material Matters, 3(2), 38-38 (2008)
Bo-Soon Kim et al.
Journal of nanoscience and nanotechnology, 13(5), 3622-3626 (2013-07-19)
A subwavelength structure (SWS) was formed via a simple chemical wet etching using a gold (Au) catalyst. Single nano-sized Au particles were fabricated by metallic self-aggregation. The deposition and thermal annealing of the thin metallic film were carried out. Thermal
Chengyong Li et al.
Journal of nanoscience and nanotechnology, 13(3), 2272-2275 (2013-06-13)
Mesoporous Si-C-O fibers were fabricated by air activation of a kind of carbon-rich SiC-C fibers at 600 degrees C. The SiC-C fibers were prepared from the hybrid precursor of polycarbosilane and pitch through melt-spinning, air curing and pyrolysis in nitrogen.
Min Joon Huang et al.
Journal of nanoscience and nanotechnology, 13(6), 3810-3817 (2013-07-19)
In this work, we demonstrated a silicon nanowire (SiNW) biosensing platform capable of simultaneously identifying different Dengue serotypes on a single sensing chip. Four peptide nucleic acids (PNAs), specific to each Dengue serotypes (DENV-1 to DENV-4), were spotted on different
Jaewoo Lee et al.
Journal of nanoscience and nanotechnology, 13(5), 3495-3499 (2013-07-19)
A spin-casting process for fabricating polycrystalline silicon sheets for use as solar cell wafers is proposed, and the parameters that control the sheet thickness are investigated. A numerical study of the fluidity of molten silicon indicates that the formation of

Articles

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

See All

Protocols

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service