Skip to Content
Merck
All Photos(1)

Key Documents

634867

Sigma-Aldrich

Silicon dioxide

single crystal substrate, optical grade, 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm

Synonym(s):

Cristobalite, Quartz, Sand, Silica

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

grade

optical grade

Assay

99.99% trace metals basis

form

solid

hardness

7.0 mohs

L × W × thickness

10 mm × 10 mm × 0.5 mm

refractive index

n20/D 1.544 (lit.)

mp

1610 °C (lit.)

transition temp

Tm 573.1 °C (phase change)

density

2.6 g/mL at 25 °C (lit.)
2.684 g/mL

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

SiO2 is majorly used as a substrate material with excellent thermo-mechanical properties in a variety of applications which include:
  • vapour deposition
  • phase deposition
  • atomic force microscopy probes(AFM)
  • spin coating
  • electronic based devices

Features and Benefits

An excellent substrate for microwave filters for applications in the wireless communication arena.

Physical form

Crystalline, hexagonal (a = 4.914 Å, c = 5.405 Å)

Other Notes

specific heat = 0.18 cal/g; thermoelectric constant = 1200μV/°C @ 300 °C; thermal conductivity = 0.0033 cal/cm°C; themal expansion (x10-6) = α11: 13.71, α33: 7.48; Q value = 1.8 × 106 min; acoustic velocity, SAW = 3160 m/sec; frequency constant, BAW = 1661 kHz/mm; piezoelectric coupling = K2 BAW: 0.65%, SAW: 0.14%

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exciton diffusion measurements in poly (3-hexylthiophene).
Shaw PE, et al.
Advanced Materials, 20(18), 3516-3520 (2008)
The effect of calcination temperature on the surface microstructure and photocatalytic activity of TiO2 thin films prepared by liquid phase deposition.
Yu J, et al.
The Journal of Physical Chemistry B, 107(50), 13871-13879 (2003)
Electrical and optical properties of ZnO transparent conducting films by the sol-gel method.
Lee J, et al.
Journal of Crystal Growth, 247(1-2), 119-125 (2003)
Growth model of bamboo-shaped carbon nanotubes by thermal chemical vapor deposition.
Lee CJ and Park J
Applied Physics Letters, 77(21), 3397-3399 (2000)
Oxygen-aided synthesis of polycrystalline graphene on silicon dioxide substrates.
Chen J, et al.
Journal of the American Chemical Society, 133(44), 17548-17551 (2011)

Articles

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service