Skip to Content
Merck
All Photos(1)

Key Documents

463035

Sigma-Aldrich

Methane

electronic grade, ≥99.998%

Synonym(s):

Marsh gas, Methyl hydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4
CAS Number:
Molecular Weight:
16.04
Beilstein:
1718732
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

vapor density

0.55 (vs air)

Assay

≥99.998%

form

gas

autoignition temp.

998 °F

expl. lim.

15 %

impurities

<2 ppm CO
<2 ppm CO2
<3 ppm Water (H2O)
<5 ppm Ethane (C2H5)
<5 ppm Nitrogen (N2)

bp

−161 °C (lit.)

mp

−183 °C (lit.)

transition temp

critical temperature −82.1 °C

density

0.716 g/mL at 25 °C (lit.)

SMILES string

[H]C([H])([H])[H]

InChI

1S/CH4/h1H4

InChI key

VNWKTOKETHGBQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Methane is used as a carbon source for the chemical vapor deposition (CVD) of carbide and diamond thin films.

Recommended products

High-purity regulators Z527378 or Z527386 are recommended.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-306.4 °F - closed cup

Flash Point(C)

-188 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceri Hammond et al.
ChemSusChem, 5(9), 1668-1686 (2012-08-01)
The economically viable oxidative upgrading of methane presents one of the most difficult but rewarding challenges within catalysis research. Its potential to revolutionalise the chemical value chain, coupled with the associated supremely challenging scientific aspects, has ensured this topic's high
Jeanette Ringvoll et al.
The EMBO journal, 25(10), 2189-2198 (2006-04-28)
Two human homologs of the Escherichia coli AlkB protein, denoted hABH2 and hABH3, were recently shown to directly reverse 1-methyladenine (1meA) and 3-methylcytosine (3meC) damages in DNA. We demonstrate that mice lacking functional mABH2 or mABH3 genes, or both, are
Judlyn Fernandes et al.
The Journal of nutrition, 143(8), 1269-1275 (2013-06-07)
Recent attention has focused on the significance of colonic Archaea in human health and energy metabolism. The main objectives of this study were to determine the associations among the number of fecal Archaea, body mass index (BMI), fecal short chain
Cornelia Welte et al.
Biochimica et biophysica acta, 1837(7), 1130-1147 (2013-12-18)
Methane-forming archaea are strictly anaerobic microbes and are essential for global carbon fluxes since they perform the terminal step in breakdown of organic matter in the absence of oxygen. Major part of methane produced in nature derives from the methyl
C Berndt et al.
Science (New York, N.Y.), 343(6168), 284-287 (2014-01-05)
Methane hydrate is an icelike substance that is stable at high pressure and low temperature in continental margin sediments. Since the discovery of a large number of gas flares at the landward termination of the gas hydrate stability zone off

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service