Skip to Content
Merck
All Photos(1)

Key Documents

295302

Sigma-Aldrich

Ethane

≥99%

Synonym(s):

Ethane gas

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH3
CAS Number:
Molecular Weight:
30.07
Beilstein:
1730716
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.05 (vs air)

Quality Level

vapor pressure

37.95 atm ( 21.1 °C)

Assay

≥99%

form

gas

autoignition temp.

881 °F

expl. lim.

13 %

bp

−88 °C (lit.)

mp

−172 °C (lit.)

density

0.362 g/mL at 20 °C (lit.)

SMILES string

CC

InChI

1S/C2H6/c1-2/h1-2H3

InChI key

OTMSDBZUPAUEDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethane, an aliphatic hydrocarbon, is a major constituent of natural and shale gas. It is a major feedstock for the preparation of ethylene and other value-added chemicals like aromatic hydrocarbons.

Application

Ethane can be used to prepare:
  • Ethylene via oxidative dehydrogenation reaction using different catalytic systems.
  • Ethanol via oxidation reaction, which is catalyzed by cytochrome P450 BM3.
  • Carbon nanofibers via decomposition method using nickel catalyst supported by a carbon nanotube.

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-211.0 °F - closed cup

Flash Point(C)

-135 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Direct conversion of ethane to ethanol by engineered cytochrome P450 BM3
Meinhold P, et al.
Chembiochem, 6(10), 1765-1768 (2005)
Catalytic Direct Conversion of Ethane to Value-added Chemicals Under Microwave Irradiation
Robinson B, et al.
Catalysis Today (2020)
Ethane to aromatic hydrocarbons: past, present, future
Hagen A and Roessner F
Catalysis Reviews: Science and Engineering, 42(4), 403-437 (2000)
Progress and prospects in catalytic ethane aromatization
Xiang Y, et al.
Catalysis Science & Technology, 8(6), 1500-1516 (2018)
Dehydrogenation of ethane with carbon dioxide over supported chromium oxide catalysts
Wang S, et al.
Applied Catalysis A: General, 196(1), 1-8 (2000)

Protocols

Separation of Methane; Acetylene; Carbon monoxide; Water; Nitrogen; Carbon dioxide; Ethane; Ethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service