Skip to Content
Merck
All Photos(1)

Key Documents

295655

Sigma-Aldrich

Propane

98%

Synonym(s):

n-Propane, Propane gas

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH3
CAS Number:
Molecular Weight:
44.10
Beilstein:
1730718
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.5 (vs air)

Quality Level

vapor pressure

190 psi ( 37.7 °C)
8.42 atm ( 21.1 °C)

Assay

98%

autoignition temp.

842 °F

expl. lim.

9.5 %

bp

-42.1 °C (lit.)

mp

-188 °C (lit.)

density

0.564 g/mL at 20 °C (lit.)

SMILES string

CCC

InChI

1S/C3H8/c1-3-2/h3H2,1-2H3

InChI key

ATUOYWHBWRKTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Propane can be used:
  • To prepare acrolein, an important raw material for the preparation of acrylic acid.
  • To prepare propene through dehydrogenation reaction.
  • As an alkylating agent for benzene.

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-155.2 °F - closed cup

Flash Point(C)

-104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Wise et al.
Journal of chemical ecology, 38(12), 1521-1527 (2012-11-28)
Bacillus subtilis strains are known to produce a vast array of antimicrobial compounds. However, some compounds remain to be identified. Disk assays performed in vitro with Bacillus subtilis CU12 showed a significant reduction in mycelial growth of Alternaria solani, Botrytis
W Ashraf et al.
FEMS microbiology letters, 122(1-2), 1-6 (1994-09-15)
The bacterial metabolism of propane and the pathway(s) involved are poorly understood, as the relative importance of terminal versus subterminal oxidation of propane, via propan-1-ol and propan-2-ol, respectively, is still unclear. In the case of bacteria, the ability to oxidize
Walker R Chan et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(14), 5309-5314 (2013-02-27)
The challenging problem of ultra-high-energy-density, high-efficiency, and small-scale portable power generation is addressed here using a distinctive thermophotovoltaic energy conversion mechanism and chip-based system design, which we name the microthermophotovoltaic (μTPV) generator. The approach is predicted to be capable of
Kil-Nam Kim et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 53, 371-375 (2012-12-26)
To assess the potential therapeutic properties of trans-1,3-diphenyl-2,3-epoxypropane-1-one (DPEP), its anti-inflammatory effects were investigated in lipopolysaccharide (LPS)-stimulated mouse macrophage (RAW 264.7) cells. DPEP induced dose-dependent reduction of the protein levels of inducible nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) and
Michał H Jamróz
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 114, 220-230 (2013-06-20)
The principle of operations of the VEDA program written by the author for Potential Energy Distribution (PED) analysis of theoretical vibrational spectra is described. Nowadays, the PED analysis is indispensible tool in serious analysis of the vibrational spectra. To perform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service