Skip to Content
Merck
All Photos(1)

Key Documents

295116

Sigma-Aldrich

Carbon monoxide

Carbon monoxide
1 of 1 reviewers received a sample product or took part in a promotion

≥99.0%

Synonym(s):

Carbon oxide (CO)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CO
CAS Number:
Molecular Weight:
28.01
Beilstein:
3587264
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

0.97 (vs air)

Quality Level

Assay

≥99.0%

autoignition temp.

1128 °F

expl. lim.

74 %

bp

−191.5 °C (lit.)

mp

−205 °C (lit.)

SMILES string

[C-]#[O+]

InChI

1S/CO/c1-2

InChI key

UGFAIRIUMAVXCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carbon monoxide is a versatile building block used in carbonylation reactions and to prepare single-walled carbon nanotubes. [1]

Application

Carbon monoxide can be used in the following applications:
  • Transition-metal-catalyzed carbonylation reactions.[2]
  • Insertion of carbon monoxide in metal-carbon bonds during copolymerization reactions.[3]
  • To grow single-wall carbon nanotubes (SWNT) via disproportionation reaction catalyzed by molybdenum particles.[4]

Catalytic hydrogenation of carbon monoxide over transition metals catalysts to form hydrocarbons has been reported.[5]

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Gas 1A - Press. Gas Compr. Gas - Repr. 1A - STOT RE 1 Inhalation

Target Organs

Heart

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa Knauert et al.
Oxidative medicine and cellular longevity, 2013, 360815-360815 (2013-01-01)
Heme oxygenase-1 (HO-1) is a regulated enzyme induced in multiple stress states. Carbon monoxide (CO) is a product of HO catalysis of heme. In many circumstances, CO appears to functionally replace HO-1, and CO is known to have endogenous anti-inflammatory
Sinisa Stanic et al.
International journal of radiation oncology, biology, physics, 88(5), 1092-1099 (2014-03-26)
To investigate pulmonary function test (PFT) results and arterial blood gas changes (complete PFT) following stereotactic body radiation therapy (SBRT) and to see whether baseline PFT correlates with lung toxicity and overall survival in medically inoperable patients receiving SBRT for
Jukka-Pekka Jalkanen et al.
Ambio, 43(3), 311-324 (2013-03-13)
This study addresses the exhaust emissions of CO₂, NO(x), SO(x), CO, and PM(2.5) originated from Baltic Sea shipping in 2006-2009. Numerical results have been computed using the Ship Traffic Emissions Assessment Model. This model is based on the messages of
Saskia Gauthier et al.
The Annals of occupational hygiene, 56(7), 755-763 (2012-08-11)
The installation of wood pellet heating as a cost-effective and climatically neutral source of energy for private households has increased steadily in recent years. We report two deaths that occurred within the space of about a year in wood pellet
Stefan W Ryter et al.
The Korean journal of internal medicine, 28(2), 123-140 (2013-03-26)
Gaseous molecules continue to hold new promise in molecular medicine as experimental and clinical therapeutics. The low molecular weight gas carbon monoxide (CO), and similar gaseous molecules (e.g., H2S, nitric oxide) have been implicated as potential inhalation therapies in inflammatory

Articles

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Protocols

Separation of Methane; Acetylene; Carbon monoxide; Water; Nitrogen; Carbon dioxide; Ethane; Ethylene

Reviews

Active Filters

  1. Cambridge
    • Reviews 7
    • Votes 15
    5 out of 5 stars.

    Carbon monoxide

    The carbon monoxide cylinder was properly protected.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service