Skip to Content
Merck
All Photos(1)

Key Documents

444685

Sigma-Aldrich

Platinum

wire, diam. 2.0 mm, 99.9% trace metals basis

Synonym(s):

Platinum black, Platinum element, Platinum gray

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
EC Number:
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

wire

resistivity

10.6 μΩ-cm, 20°C

diam.

2.0 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

Platinum (Pt) wire is a metal wire that is a biocompatible material that can be used in electrodes and implants. The efficiency of the platinum electrode can be improved by forming a platinum black surface.

Quantity

1.4 g = 2 cm; 7 g = 10 cm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrodes
Bioimpedance and Bioelectricity Basics, 179-254 (2015)
Synthesis of novel ruthenium sensitizers and their application in dye-sensitized solar cells
Nazeeruddin MK, et al.
Coordination Chemistry Reviews, 249(13-14), 1460-1467 (2005)
Electrode array design and fabrication for implantable systems
Implantable Sensor Systems for Medical Applications, 249(13-14), 150-182 (2013)
Wei Sun et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 1907-1913 (2013-03-19)
In this paper a platinum (Pt) nanoparticle decorated graphene (GR) nanosheet was synthesized and used for the investigation on direct electrochemistry of myoglobin (Mb). By integrating GR-Pt nanocomposite with Mb on the surface of carbon ionic liquid electrode (CILE), a
Shai Rozen et al.
Plastic and reconstructive surgery, 131(6), 1253-1265 (2013-05-30)
Left untreated, paralytic lagophthalmos may result in corneal dryness, ulcerations, and subsequent blindness. The most common nondynamic surgical solution is upper eyelid weight placement in a superficial, pretarsal pocket, carrying the risk of visibility, extrusion, and entropion. The authors present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service