Skip to Content
Merck
All Photos(5)

Key Documents

205273

Sigma-Aldrich

Titanium(IV) isopropoxide

97%

Synonym(s):

TTIP, Tetraisopropyl orthotitanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti[OCH(CH3)2]4
CAS Number:
Molecular Weight:
284.22
Beilstein:
3679474
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

reaction suitability

core: titanium
reagent type: catalyst

refractive index

n20/D 1.464 (lit.)

bp

232 °C (lit.)

mp

14-17 °C (lit.)

density

0.96 g/mL at 20 °C (lit.)

SMILES string

CC(C)O[Ti](OC(C)C)(OC(C)C)OC(C)C

InChI

1S/4C3H7O.Ti/c4*1-3(2)4;/h4*3H,1-2H3;/q4*-1;+4

InChI key

VXUYXOFXAQZZMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV) isopropoxide is a titanium-based coordination compound, commonly used in the asymmetric Sharpless epoxidation reaction of allylic alcohols. It is also used as a catalyst in Kulinkovich reaction for the synthesis of cyclopropanes.

Application

Commonly used as a precursor for the preparation of Titania (TiO2)
Novel metal oxide/phosphonate hybrids were formed from titanium(IV) isopropoxide in a two-step sol-gel process. Starting material for barium-strontium-titanate thin films. Used to make porous titanosilicates, potential ion-exchange materials for cleanup of radioactive wastes. Applied in the formation of a heterosupermolecule consisting of a TiO2 nanocrystallite-viologen electron acceptor complex whose light-induced electron transfer has been demonstrated.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Product of Dorf Ketal
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

105.8 °F - Pensky-Martens closed cup

Flash Point(C)

41 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric epoxidation of allylic alcohols: the Katsuki-Sharpless epoxidation reaction
Katsuki T and Martin V
Organic Reactions, 48(3), 1-299 (2004)
Al-Jufairi, N.
Materials Science Forum, 517, 165-172 (2006)
J A Darr et al.
IEE proceedings. Nanobiotechnology, 152(3), 109-111 (2006-01-31)
Surfactant-coated amorphous titania nanospheres have been synthesised using templating 'water-in-supercritical carbon dioxide' emulsion droplets; the process represents a clean and controlled method for the manufacture of high-purity nanoparticles.
Ethers, Epoxides, and Sulfides
Organic Chemistry (2008)
Titanium (IV) isopropoxide-catalyzed formation of 1-substituted cyclopropanols in the reaction of ethylmagnesium bromide with methyl alkanecarboxylates
Kulinkovich OG, et al.
Synthesis, 3, 234-234 (1991)

Articles

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service