Skip to Content
Merck
All Photos(1)

Key Documents

327492

Sigma-Aldrich

Platinum

wire, diam. 1.0 mm, 99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
EC Number:
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

wire

resistivity

10.6 μΩ-cm, 20°C

diam.

1.0 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Related Categories

General description

This platinum wire is a thin cylindrical rod of platinum with a diameter of 1 mm. Platinum is a dense, silver-white metal that is highly resistant to corrosion and has a high melting point. Platinum is a precious metal that is valued for its rarity, chemical stability, and catalytic properties. Platinum wire is often used as electrodes in fuel cells, batteries, and other electrochemical devices, in catalytic converters in the automotive industry, in medical implants, and in heating elements and thermocouples.

Application

Platinum wire may be used as an electrode during various electrochemical studies. It may also be used during the solid state deposition colorless transparent crystals of a new noncentrosymmetric hexaborate, KBOC, on it surface. Platinum wire, as a catalyst may be useful in accelerating the generation of hydrogen from Ni-rich magnesium alloy in NaCl solution.

Quantity

520 mg = 30 mm; 2.6 g = 150 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K3B6O10Cl: a new structure analogous to perovskite with a large second harmonic generation response and deep UV absorption edge
Wu H, et al.
Journal of the American Chemical Society, 133(20), 7786-7790 (2011)
Wire-like charge transport at near constant bridge energy through fluorene oligomers.
Goldsmith RH, et al.
Proceedings of the National Academy of Sciences of the USA, 102(10), 3540-3545 (2005)
K3B6O10Cl: a new structure analogous to perovskite with a large second harmonic generation response and deep UV absorption edge.
Wu H, et al.
Journal of the American Chemical Society, 133(20), 7786-7790 (2011)
Evaluation of a new hydrogen generating system: Ni-rich magnesium alloy catalyzed by platinum wire in sodium chloride solution.
Cho CY, et al.
Materials Transactions, 46(12), 2704-2708 (2005)
Wei Sun et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 1907-1913 (2013-03-19)
In this paper a platinum (Pt) nanoparticle decorated graphene (GR) nanosheet was synthesized and used for the investigation on direct electrochemistry of myoglobin (Mb). By integrating GR-Pt nanocomposite with Mb on the surface of carbon ionic liquid electrode (CILE), a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service