Skip to Content
Merck
All Photos(1)

Key Documents

349402

Sigma-Aldrich

Platinum

wire, diam. 0.25 mm, 99.9% trace metals basis

Synonym(s):

Platinum black, Platinum element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
EC Number:
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

wire

resistivity

10.6 μΩ-cm, 20°C

diam.

0.25 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

Platinum (Pt) based wire is a dense, silvery metal that is highly resistant to corrosion. It is a soft transition metal that is highly ductile and malleable, which makes it useful in electronic industries. It has a high melting point and can be oxidized at 500°C.

Application

Pt wires can be used for a wide range of applications such as:
  • reference electrodes for electrochemical processes
  • for the formation of microchannels for microfluidic devices
  • coiling of an optical fiber
  • development of cathode for electrolyte based fuel cells

Quantity

250 mg = 25 cm; 1.5 g = 150 cm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effect of voltage on platinum dissolution relevance to polymer electrolyte fuel cells
Wang X, et al.
Electrochemical and Solid-State Letters, 9(5), A225-A227 (2006)
Platinum Metals Rev. (2004)
Electrochemistry of europium (III) chloride in 3 LiCl-NaCl, 3 LiCl-2 KCl, LiCl-RbCl, and 3 LiCl-2 CsCl eutectics at various temperatures
Schroll CA, et al.
Journal of the Electrochemical Society, 164(8), H5345-H5352 (2017)
Characterizing Electro-osmotic Flow in Parylene Microchannels
Freire SLS, et al.
Polymer-Plastics Technology and Engineering, 50(9), 931-936 (2011)
Chemistry Foundations and Applications (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service