Skip to Content
Merck
All Photos(2)

Key Documents

204315

Sigma-Aldrich

Selenium dioxide

99.999% trace metals basis

Synonym(s):

NSC 56753, Selenium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SeO2
CAS Number:
Molecular Weight:
110.96
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 157 °C)

Assay

99.999% trace metals basis

form

solid

mp

315 °C (subl.) (lit.)

SMILES string

O=[Se]=O

InChI

1S/O2Se/c1-3-2

InChI key

JPJALAQPGMAKDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mild oxidizing reagent for allylic olefins and acetylenes. Conversion of 1,3-diketones to 1,2,3-triones.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 34, 2979-2979 (1993)
Asian Journal of Chemistry, 19, 4107-4107 (2007)
Vijay P Singh et al.
Chemistry, an Asian journal, 6(6), 1431-1442 (2011-05-11)
The syntheses of selenenate/seleninate esters and related derivatives by aromatic nucleophilic substitution (S(N)Ar) reactions of 2-bromo-3-nitrobenzylalcohol (13) and 2-bromo-3-nitrobenzaldehyde (17) with Na(2)Se(2)/nBuSeNa are described. The reaction of 13 with Na(2)Se(2) at room temperature afforded 7-nitro-1,2-benzisoselenole(3H) (15) instead of the desired
Zhangrong Lou et al.
Chemical communications (Cambridge, England), 49(24), 2445-2447 (2013-02-19)
Based on a novel strategy for modulating the fluorescence of selenide and selenoxide, we have designed and developed a reversible fluorescent probe for hypochloric acid. And the synthesis, characterization, fluorescence properties, as well as the biological applications in living cells
Zoyne Pedrero et al.
Journal of chromatography. A, 1139(2), 247-253 (2006-12-05)
Selenium speciation in Se-enriched Lens esculenta grown in hydroponic culture containing inorganic selenium as Na(2)SeO(3) and Na(2)SeO(4) was performed. After 16 days of growth, the plants were collected and divided in two parts, roots and stems and then analysed to

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service