Skip to Content
Merck
All Photos(1)

Key Documents

142433

Sigma-Aldrich

Phenylglyoxal hydrate

97%

Synonym(s):

2,2-Dihydroxyacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCHO · xH2O
CAS Number:
Molecular Weight:
134.13 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

bp

142 °C/125 mmHg (lit.)

mp

76-79 °C (lit.)

solubility

95% ethanol: soluble 5%, clear to very slightly hazy, colorless to light yellow

functional group

aldehyde
ketone
phenyl

SMILES string

[H]O[H].[H]C(=O)C(=O)c1ccccc1

InChI

1S/C8H6O2.H2O/c9-6-8(10)7-4-2-1-3-5-7;/h1-6H;1H2

InChI key

YQBLQKZERMAVDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenylglyoxal hydrate was used :
  • to modify pig muscle carbonic anhydraseIII
  • as specific reagent for arginine groups
  • to prepare pyrrolinone and furan derivatives
  • as chemiluminescent reagent for the determination of purines

Biochem/physiol Actions

Phenylglyoxal is a potent inhibitor of mitochondrial aldehyde dehydrogenase. It reacts with arginine residues in purified Hageman factor (HF, Factor XII) and causes inhibition of its coagulant properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

O D Radnoff et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 148(1), 177-182 (1975-01-01)
Exposure of purified Hageman factor (HF, Factor XII) to phenylglyoxal hydrate (PHG), an agent reacting with arginine residues in protein, inhibited its coagulant properties upon subsequent exposure of negatively charged agents. Once HF had been exposed to kaolin or ellagic
L M Pullan et al.
Biochemistry, 24(3), 635-640 (1985-01-29)
Mammalian carbonic anhydrase III has previously been shown to catalyze the hydrolysis of p-nitrophenyl phosphate in addition to possessing the conventional CO2 hydratase and p-nitrophenylacetate esterase activities. Modification of pig muscle carbonic anhydrase III with the arginine reagent phenylglyoxal yielded
Synthesis, 783-783 (1993)
William A Irwin et al.
Biochemical and biophysical research communications, 291(2), 215-219 (2002-02-16)
Fructose has been shown to protect hepatocyte viability during hypoxia or exposure to mitochondrial electron transport inhibitors. We report here that the fructose metabolite D-glyceraldehyde (D-GA) is a good inhibitor of the mitochondrial permeability transition pore (PTP) in isolated rat
Analytica Chimica Acta, 278, 275-275 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service