Skip to Content
Merck
All Photos(1)

Key Documents

180629

Sigma-Aldrich

Diphenyl diselenide

98%

Synonym(s):

Phenyl diselenide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SeSeC6H5
CAS Number:
Molecular Weight:
312.13
Beilstein:
2047179
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

impurities

<2% diphenyl selenide

mp

59-61 °C (lit.)

SMILES string

[Se]([Se]c1ccccc1)c2ccccc2

InChI

1S/C12H10Se2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H

InChI key

YWWZCHLUQSHMCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diphenyl Diselenide is an organo selenium compound that is an oxidized derivative of benzeneselenol. It is a source of phenylseleno radicals. it is reactive with a wide range of organic nucleophiles, electrophiles, and radicals.

Application

Diphenyl diselenide is used:In the synthesis of diaryl selenides bycross-coupling of aryl halides with diphenyl diselenide in presence of copperferrite nanoparticle catalyst.

Biochem/physiol Actions

Diphenyl diselenide inhibits δ-aminolevulinate dehydratase (δ-ALA-D) from brain, liver and kidney in vitro.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E N Maciel et al.
Journal of biochemical and molecular toxicology, 14(6), 310-319 (2000-11-18)
In the present study, the inhibitory effect of diphenyl diselenide and diphenyl ditelluride after in vitro, acute (a single dose), or chronic exposure (14 doses) was examined in mice 24 hours after the last administration. In vitro, diphenyl diselenide, and
Diphenyl Diselenide-Catalyzed Synthesis of Triaryl Phosphites and Triaryl Phosphates from White Phosphorus
Zhang Y, et al.
Organic Letters, 23(13), 5158-5163 (2021)
Diphenyl Diselenide
Byers JH
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Pengshang Zhou et al.
Nanoscale, 12(43), 22307-22316 (2020-11-05)
Transition metal dichalcogenides (TMDs) are gaining increasing interest in the field of lithium ion batteries due to their unique structure. However, previous preparation methods have mainly focused on their growth from substrates or by exfoliation of the bulk materials. Considering
Erico Silva Loreto et al.
Veterinary microbiology, 156(1-2), 222-226 (2011-11-08)
This study evaluated the in vitro activity of diphenyl diselenide against 19 Pythium insidiosum isolates and the in vivo therapeutic response of rabbits with experimentally induced pythiosis. In vitro: susceptibility tests were performed using the broth macrodilution method in accordance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service