Skip to Content
Merck
All Photos(4)

Key Documents

325473

Sigma-Aldrich

Selenium dioxide

reagent grade, powder, 98%

Synonym(s):

NSC 56753, Selenium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SeO2
CAS Number:
Molecular Weight:
110.96
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

vapor pressure

1 mmHg ( 157 °C)

Assay

98%

form

powder

reaction suitability

reagent type: oxidant

color

off-white to pink

mp

315 °C (subl.) (lit.)

SMILES string

O=[Se]=O

InChI

1S/O2Se/c1-3-2

InChI key

JPJALAQPGMAKDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Selenium dioxide (SeO2) is mainly used to oxidize the α-methylene carbon atoms adjacent to a double bond to form allylic hydroxy derivatives.
Other applications:
  • SeO2 reacts with olefins in the presence of tert-butyl hydrogen peroxide to form allylic alcohols without the formation of rearrangement or dehydration by-products. This reagent system is also useful for the oxidation reactions of acetylenic fatty esters.
  • SeO2 can be used for the oxidation of 1,3,5-cycloheptatriene to form tropone.
  • Piperidoindole systems on oxidation with SeO2 forms either 2-acyl- or 3-acyl-indoles or fully aromatic β-carbolines, depending on the starting material.
  • SeO2 reacts with chlorotrimethylsilane to form selenium oxychloride in situ, which is an effective chlorinating reagent for alcohols. This reagent is a better alternative to thionyl chloride and other chlorinating agents.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation reactions of acetylenic fatty esters with selenium dioxide/tert-butyl hydroperoxide.
Jie M S L K, et al.
Lipids, 32(10), 1119-1123 (1997)
Tropone. Selenium Dioxide Oxidation of 1, 3, 5-Cycloheptatriene.
Radlick P.
The Journal of Organic Chemistry, 29(4), 960-960 (1964)
Selenium dioxide oxidations in the indole area. Synthesis of β-carboline alkaloids.
Cain M, et al.
Journal of the American Chemical Society, 105(4), 907-913 (1983)
Tao Huang et al.
International journal of nanomedicine, 15, 4275-4288 (2020-07-02)
Selenium nanoparticles (Se NPs) are promising antibacterial agents to tackle the growing problem of antimicrobial resistance. The aim of this study was to fabricate Se NPs with a net positive charge to enhance their antibacterial efficacy. Se NPs were coated
Allylic oxidation of olefins by catalytic and stoichiometric selenium dioxide with tert-butyl hydroperoxide.
Umbreit M A and Sharpless K B
Journal of the American Chemical Society, 99(16), 5526-5528 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service