Skip to Content
Merck
All Photos(1)

Key Documents

131490

Sigma-Aldrich

4-Methylpyridine

98%

Synonym(s):

4-Picoline, 4-Methylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
Beilstein:
104586
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.2 (vs air)

Quality Level

vapor pressure

4 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

1000 °F

refractive index

n20/D 1.504 (lit.)

bp

145 °C (lit.)

mp

2.4 °C (lit.)

density

0.957 g/mL at 25 °C (lit.)

SMILES string

Cc1ccncc1

InChI

1S/C6H7N/c1-6-2-4-7-5-3-6/h2-5H,1H3

InChI key

FKNQCJSGGFJEIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methylpyridine is a neutral N-donor ligand.

4-picoline (4-Mepy) forms isostructural complexes with iron and cobalt phthalocyanines.

Application

4-Methylpyridine was used in the preparation of a 1,2-dihydropyridide derivative.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative study of beryllium, magnesium and zinc phthalocyanine complexes with 4-picoline
Kubiak R, et al.
Polyhedron, 26, 4179-4186 (2007)
Susan M Chemaly et al.
Inorganic chemistry, 52(2), 1077-1083 (2012-12-28)
Equilibrium constants (log K) for substitution of coordinated H(2)O in aquacyanocobyrinic acid heptamethyl ester (aquacyanocobester, ACCbs) and aquacyano-stable yellow cobyrinic acid hexamethyl ester (aquacyano-stable yellow cobester, ACSYCbs), in which oxidation of the C5 carbon of the corrin interrupts the normal
Michael S Hill et al.
Dalton transactions (Cambridge, England : 2003), 40(46), 12500-12509 (2011-10-12)
Reactions of the β-diketiminato n-butyl magnesium complex, [HC{(Me)CN(2,6-(i)Pr(2)C(6)H(3))}(2)Mg(n)Bu], with a range of substituted pyridines and fused-ring quinolines in the presence of PhSiH(3) has been found to result in dearomatisation of the N-heterocyclic compounds. This reaction is proposed to occur through
Stéphanie Romand et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 63, 14-21 (2014-07-06)
A capillary isoelectric focusing (cIEF) method was developed for the determination of the ionization constants (pKa) of small molecules. Two approaches used to decrease the electroosmotic flow (EOF) were compared: (i) a hydroxypropylcellulose (HPC) coated capillary in aqueous medium and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service